Merck
  • Trypanosoma congolense Infections: Induced Nitric Oxide Inhibits Parasite Growth In Vivo.
Trypanosoma congolense Infections: Induced Nitric Oxide Inhibits Parasite Growth In Vivo.
Journal of parasitology research (2011-05-18)
Wenfa Lu, Guojian Wei, Wanling Pan, Henry Tabel
ABSTRACT
Wild-type (WT) C57BL/6 mice infected intraperitoneally with 5 × 10(6) Trypanosoma congolense survive for more than 30 days. C57BL/6 mice deficient in inducible nitric oxide synthase (iNOS(-/-)) and infected with 10(3) or 5 × 10(6) parasites do not control the parasitemia and survive for only 14 ± 7 or 6.8 ± 0.1 days, respectively. Bloodstream trypanosomes of iNOS(-/-) mice infected with 5 × 10(6)T. congolense had a significantly higher ratio of organisms in the S+G2+M phases of the cell cycle than trypanosomes in WT mice. We have reported that IgM anti-VSG-mediated phagocytosis of T. congolense by macrophages inhibits nitric oxide (NO) synthesis via CR3 (CD11b/CD18). Here, we show that during the first parasitemia, but not at later stages of infection, T. congolense-infected CD11b(-/-) mice produce more NO and have a significantly lower parasitemia than infected WT mice. We conclude that induced NO contributes to the control of parasitemia by inhibiting the growth of the trypanosomes.
Product Description
Supelco
Sulfanilamide melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sulfanilamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, puriss., ≥99%
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O