Skip to Content
Merck
All Photos(1)

Key Documents

30417

Sigma-Aldrich

Phosphoric acid

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%

Synonym(s):

Orthophosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3PO4
CAS Number:
Molecular Weight:
98.00
Beilstein:
1921286
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
Agency:
USP/NF
reag. ISO
reag. Ph. Eur.
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor density

3.4 (vs air)

vapor pressure

2.2 mmHg ( 20 °C)
5 mmHg ( 25 °C)

Assay

≥85%

form

liquid

impurities

reducing impurities, in accordance
residual solvents, in accordance
substances precipitated by ammonia, in accordance
≤0.001% KMnO4 red. matter (as O)
≤0.001% heavy metals (as Pb)
≤0.001% in water insoluble matters
≤0.001% volatile acids (as CH3COOH)

bp

158 °C (lit.)

mp

~40 °C (lit.)

density

1.685 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤2 mg/kg
fluoride (F-): ≤1 mg/kg
nitrate (NO3-): ≤3 mg/kg
phosphite, hypophosphite (as H3PO3): ≤20 mg/kg
phosphite, hypophosphite (as H3PO3): in accordance
sulfate (SO42-): ≤30 mg/kg

cation traces

As: ≤0.5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤5 mg/kg
K: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤0.5 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sb: ≤2 mg/kg
Zn: ≤2 mg/kg

SMILES string

OP(O)(O)=O

suitability

passes test for appearance of solution

InChI

1S/H3O4P/c1-5(2,3)4/h(H3,1,2,3,4)

InChI key

NBIIXXVUZAFLBC-UHFFFAOYSA-N

Gene Information

human ... SRC(6714)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Cooperative Rh/Achiral Phosphoric Acid-Enabled [3+3] Cycloannulation of Carbonyl Ylides with Quinone Monoimines: Synthesis of Benzofused Dioxabicyclo[3.2.1]octane Scaffolds.: This study demonstrates the catalytic role of phosphoric acid in a [3+3] cycloannulation reaction, enabling the efficient synthesis of complex organic scaffolds (Tu et al., 2024).

Other Notes

Concentrated phosphoric acid is approx. 85% H3PO4 (aqueous).
The article number 30417-4X2.5L will be discontinued. Please order the single bottle 30417-2.5L which is physically identical with the same exact specifications.
The article number 30417-4X2.5L-M will be discontinued. Please order the single bottle 30417-2.5L-M which is physically identical with the same exact specifications.
The article number 30417-6X1L will be discontinued. Please order the single bottle 30417-1L which is physically identical with the same exact specifications.
The article number 30417-6X1L-M will be discontinued. Please order the single bottle 30417-1L-M which is physically identical with the same exact specifications.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Internal versus external surface active sites in ZSM-5 zeolite: Part 1. Fries rearrangement catalyzed by modified and unmodified H3PO4/ZSM-5.
Ghiaci M, et al.
Applied Catalysis A: General, 298, 32-39 (2006)
J M Fujitaki et al.
Biochemistry, 20(12), 3658-3664 (1981-06-09)
Phosphorus-31 nuclear magnetic resonance (31P NMR) is used to investigate acid-labile phosphorylation of histone H4. 31P NMR detects phosphorylated histidine residues in in vitro enzymatically phosphorylated H4. The source of kinase is nuclei from either regenerating rat liver or Walker-256
D S Marinier et al.
Journal of lipid research, 29(9), 1237-1243 (1988-09-01)
In order to study the lipid composition of human white matter, we have developed a 31P NMR spectroscopy method, which allows the determination and quantitation of the main phospholipids found in biological membranes. The technique is based upon the use
P Meneses et al.
Journal of lipid research, 29(5), 679-689 (1988-05-01)
The common phospholipids from biological sources were quantitated using phosphorus-31 nuclear magnetic resonance (NMR) spectroscopy in conjunction with an analytical reagent composed of two parts: 1) 2 ml of reagent chloroform in which was dissolved 0.01-100 mg of crude tissue
Alessio Ciulli et al.
Journal of medicinal chemistry, 49(16), 4992-5000 (2006-08-04)
Mapping interactions at protein-ligand binding sites is an important aspect of understanding many biological reactions and a key part of drug design. In this paper, we have used a fragment-based approach to probe "hot spots" at the cofactor-binding site of

Protocols

Folic acid vs. folate differences and biological activities; implications in fortified foods and supplements for human health.

Folic acid vs. folate differences and biological activities; implications in fortified foods and supplements for human health.

Folic acid vs. folate differences and biological activities; implications in fortified foods and supplements for human health.

Folic acid vs. folate differences and biological activities; implications in fortified foods and supplements for human health.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service