Skip to Content
Merck
All Photos(1)

Key Documents

04107

Sigma-Aldrich

Phosphoric acid

puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%

Synonym(s):

Orthophosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3PO4
CAS Number:
Molecular Weight:
98.00
Beilstein:
1921286
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.4 (vs air)

Quality Level

vapor pressure

2.2 mmHg ( 20 °C)
5 mmHg ( 25 °C)

grade

puriss.

Assay

85.0-88.0%

form

liquid

quality

meets analytical specification of Ph. Eur., BP, NF, FCC

impurities

alkali phosphates, in accordance
residual solvents, in accordance
substance precip. by Ammonia, in accordance
≤0.0005% heavy metals (as Pb)
≤0.001% volatile acids (as CH3COOH)

bp

158 °C (lit.)

mp

~40 °C (lit.)

density

1.685 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
fluoride (F-): ≤5 mg/kg
nitrate (NO3-): ≤5 mg/kg
nitrate (NO3-): in accordance
phosphite, hypophosphite (as H3PO3): ≤30 mg/kg
phosphite, hypophosphite (as H3PO3): in accordance
sulfate (SO42-): ≤50 mg/kg
sulfate (SO42-): in accordance

cation traces

As: ≤2 mg/kg
Cd: ≤3 mg/kg
Cu: ≤10 mg/kg
Fe: ≤10 mg/kg
Hg: ≤1 mg/kg
Pb: ≤3 mg/kg
Zn: ≤10 mg/kg

suitability

passes test for appearance of solution

SMILES string

OP(O)(O)=O
OP(O)(O)=O

InChI

1S/H3O4P/c1-5(2,3)4/h(H3,1,2,3,4)

InChI key

NBIIXXVUZAFLBC-UHFFFAOYSA-N

Gene Information

human ... SRC(6714)

Looking for similar products? Visit Product Comparison Guide

Other Notes

Concentrated phosphoric acid is approx. 85% H3PO4 (aqueous).
The article number 04107-4X2.5L will be discontinued. Please order the single bottle 04107-2.5L which is physically identical with the same exact specifications.
The article number 04107-6X1L will be discontinued. Please order the single bottle 04107-1L which is physically identical with the same exact specifications.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Fujitaki et al.
Biochemistry, 20(12), 3658-3664 (1981-06-09)
Phosphorus-31 nuclear magnetic resonance (31P NMR) is used to investigate acid-labile phosphorylation of histone H4. 31P NMR detects phosphorylated histidine residues in in vitro enzymatically phosphorylated H4. The source of kinase is nuclei from either regenerating rat liver or Walker-256
Internal versus external surface active sites in ZSM-5 zeolite: Part 1. Fries rearrangement catalyzed by modified and unmodified H3PO4/ZSM-5.
Ghiaci M, et al.
Applied Catalysis A: General, 298, 32-39 (2006)
D S Marinier et al.
Journal of lipid research, 29(9), 1237-1243 (1988-09-01)
In order to study the lipid composition of human white matter, we have developed a 31P NMR spectroscopy method, which allows the determination and quantitation of the main phospholipids found in biological membranes. The technique is based upon the use
P Meneses et al.
Journal of lipid research, 29(5), 679-689 (1988-05-01)
The common phospholipids from biological sources were quantitated using phosphorus-31 nuclear magnetic resonance (NMR) spectroscopy in conjunction with an analytical reagent composed of two parts: 1) 2 ml of reagent chloroform in which was dissolved 0.01-100 mg of crude tissue
Li Yan Chan et al.
The Journal of organic chemistry, 78(17), 8826-8832 (2013-08-07)
A simple and efficient method is developed for Pd-catalyzed ortho-acetoxylation using organophosphates, namely, benzylic phosphonic and aryl phosphoric monoacids, as the directing group.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service