Merck
Antitubercular activity of alpha,omega-diaminoalkanes, H2N(CH2)nNH2.
Bioorganic & medicinal chemistry letters (2009-08-04)
Fátima M F Vergara, Maria das G M O Henriques, André L P Candea, Jim L Wardell, Marcus V N De Souza
ABSTRACT
A series of 11 alpha,omega-diaminoalkanes, (H(2)N(CH(2))(n)NH(2), n=2-12) have been evaluated for their in vitro antibacterial activity against Mycobacterium tuberculosis H37Rv. Compounds, (H(2)N(CH(2))(n)NH(2), n=9-12), exhibited a very good activities in the range 2.50-3.12 microg/mL, which can be compared with that of the first line drug, ethambutol (3.12 microg/mL). These results and a preliminary QSAR study can be considered an important start point for the rational design of new leads for anti-TB compounds.
Sigma-Aldrich
1,8-Diaminooctane, 98%
Sigma-Aldrich
1,4-Diaminobutane, 99%
Sigma-Aldrich
1,12-Diaminododecane, 98%
Sigma-Aldrich
Cadaverine, 95%
Sigma-Aldrich
1,7-Diaminoheptane, 98%
Sigma-Aldrich
Hexamethylenediamine, technical grade, 70%
Sigma-Aldrich
1,9-Diaminononane, 98%
Sigma-Aldrich
Ethylenediamine, BioXtra
Sigma-Aldrich
Ethylenediamine, meets USP testing specifications
Sigma-Aldrich
1,10-Diaminodecane, 97%
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%
Supelco
Ethylenediamine, analytical standard
Sigma-Aldrich
Ethylenediamine solution, technical, 75-80%
Sigma-Aldrich
1,3-Diaminopropane, ≥99%
Sigma-Aldrich
Ethylenediamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Hexamethylenediamine, 98%
Sigma-Aldrich
Ethylenediamine, puriss. p.a., absolute, ≥99.5% (GC)