D14204
1,10-Diaminodecane
97%
Synonym(s):
1,10-Decanediamine, Decamethylenediamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
NH2(CH2)10NH2
CAS Number:
Molecular Weight:
172.31
Beilstein:
1738591
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
bp
140 °C/12 mmHg (lit.)
mp
59-61 °C (lit.)
SMILES string
NCCCCCCCCCCN
InChI
1S/C10H24N2/c11-9-7-5-3-1-2-4-6-8-10-12/h1-12H2
InChI key
YQLZOAVZWJBZSY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Sen-Ichi Aizawa et al.
Chirality, 28(1), 85-91 (2015-10-30)
The optically active mixed-ligand fac(S)-tris(thiolato)rhodium(III) complexes, ΔL -fac(S)-[Rh(aet)2 (L-cys-N,S)](-) (aet = 2-aminoethanethiolate, L-cys = L-cysteinate) () and ΔLL -fac(S)-[Rh(aet)(L-cys-N,S)2 ](2-) were newly prepared by the equatorial preference of the carboxyl group in the coordinated L-cys ligand. The amide formation reaction
Jie Zhang et al.
Journal of biotechnology, 299, 79-85 (2019-05-03)
Glucosinolates can be hydrolyzed by the enzyme commonly known as myrosinase (E.C. 3.2.1.147) to a variety of biological compounds. Myrosinase (MYR) has been immobilized through the flexible spacers of different length on cross-linked chitosan resin (CCR). Ethylene diamine, hexamethylenediamine and
Dmitry V Klinov et al.
Colloids and surfaces. B, Biointerfaces, 196, 111321-111321 (2020-08-26)
High-resolution atomic force microscopy (AFM) of biomacromolecules is a valuable method for structural studies in biology. Traditionally, the surfaces used for AFM imaging of individual molecules are limited to mica, graphite, and glass. Because these substrates have certain shortcomings, new
Gabriela E Davey et al.
Nature communications, 8(1), 1575-1575 (2017-11-18)
The 'acidic patch' is a highly electronegative cleft on the histone H2A-H2B dimer in the nucleosome. It is a fundamental motif for protein binding and chromatin dynamics, but the cellular impact of targeting this potentially therapeutic site with exogenous molecules
Lei He et al.
Scientific reports, 8(1), 767-767 (2018-01-18)
G-quadruplexes (GQ) folded by the oncogenic G-rich sequences are the promising targets for developing anticancer therapeutic molecules. However, the current drug development mainly focused on non-covalent dynamic binders to stabilize GQ structures, while the covalent targeting from inorganic complexes via
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service