Skip to Content
Merck
All Photos(2)

Key Documents

422002

Sigma-Aldrich

Hexamethylenediamine

technical grade, 70%

Synonym(s):

1,6-Diaminohexane, 1,6-Hexanediamine, HMDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)6NH2
CAS Number:
Molecular Weight:
116.20
Beilstein:
1098307
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
grade:
technical grade
Assay:
70%
technique(s):
gas chromatography (GC): suitable
bp:
111-130 °C
Pricing and availability is not currently available.

grade

technical grade

Quality Level

vapor density

4 (vs air)

Assay

70%

form

liquid

expl. lim.

6.3 %

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.439 (lit.)

bp

111-130 °C

mp

42-45 °C (lit.)

solubility

alcohol: soluble(lit.)
hydrocarbons: soluble(lit.)
water: soluble(lit.)

density

0.89 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

NCCCCCCN

InChI

1S/C6H16N2/c7-5-3-1-2-4-6-8/h1-8H2

InChI key

NAQMVNRVTILPCV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Hexamethylenediamine (1,6-Diaminohexane) is a linear aliphatic diamine. It reacts with various inorganic acids to afford mono and diacid salts. It can be prepared from adipic dinitrile via catalytic hydrogenation reaction.[1]
Hexamethylenediamine is a useful industrial intermediate widely used in the production of aliphatic and semi-aromatic polyamides.[2] Analysis of 1,6-hexamethylenediamine (HDA) in aqueous solution, human urine and plasma by capillary gas chromatographic method with thermionic specific detection has been reported.[3]

Application

Hexamethylenediamine (1,6-hexanediamine, HDA ) may be used in the preparation of the following:
  • polyamides and their hydrolytic and fungal degradability was investigated[4]
  • mullite ceramics[5]
  • dialkyl hexamethylene-1,6-dicarbamate (AHDC), via trans-esterification reaction of HDA and alkyl carbamates (AC) using FeCl3 as catalyst[6]

Other Notes

remainder water

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

201.2 °F

Flash Point(C)

94 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromatographic determination of amines in biological fluids with special reference to the biological monitoring of isocyanates and amines: I. Determination of 1, 6-hexamethylenediamine using glass capillary gas chromatography and thermionic specific detection.
Skarping G, et al.
Journal of Chromatography A, 479, 125-133 (1989)
Preparation of mullite ceramics from coprecipitated aluminum hydroxide and kaolinite using hexamethylenediamine.
Pascual J, et al.
Journal of the American Ceramic Society. American Ceramic Society, 83(11), 2677-2680 (2000)
Hydrolytic and fungal degradation of polyamides derived from tartaric acid and hexamethylenediamine.
Marques MS, et al.
Polymer, 41(8), 2765-2772 (2000)
Eagleson M.
Concise Encyclopedia Chemistry, 313-313 (1994)
The catalytic hydrogenation of adiponitrile to hexamethylenediamine over a rhodium/alumina catalyst in a three phase slurry reactor.
Alini S, et al.
J. Mol. Catal. A: Chem., 206(1), 262-270 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service