Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Twin tec PCR plates, 96 wells
Products
Product Category
plates (20)
Manufacturer
Wells
Color
Package Size
Sterility
Technique
Filter Products
Showing 1-20 of 20
Compare
Product Number
Description
Pricing
EP0030128575
Eppendorf
®
Twin tec PCR plates, 96 wells,
colorless plate, conical bottom colorless wells, semi-skirted, pkg of 25 ea
Expand
View Pricing
EP0030128648
Eppendorf
®
Twin tec PCR plates, 96 wells,
colorless plate, conical bottom colorless wells, skirted, pkg of 25 ea
Expand
View Pricing
EP0030128613
Eppendorf
®
Twin tec PCR plates, 96 wells,
red plate, conical bottom colorless wells, semi-skirted, pkg of 25 ea
Expand
View Pricing
EP0030128591
Eppendorf
®
Twin tec PCR plates, 96 wells,
green plate, conical bottom colorless wells, semi-skirted, pkg of 25 ea
Expand
View Pricing
EP0030128869
Eppendorf
®
Twin tec PCR plates, 96 wells,
colorless plate, conical bottom colorless wells, semi-skirted, pkg of 300 ea
Expand
View Pricing
EP0030128605
Eppendorf
®
Twin tec PCR plates, 96 wells,
blue plate, conical bottom colorless wells, semi-skirted, pkg of 25 ea
Expand
View Pricing
EP0030128672
Eppendorf
®
Twin tec PCR plates, 96 wells,
blue plate, conical bottom colorless wells, skirted, pkg of 25 ea
Expand
View Pricing
EP0030128770
Eppendorf
®
Twin tec PCR plates, 96 wells,
colorless plate, conical bottom colorless wells, skirted, pkg of 300 ea
Expand
View Pricing
EP0030128478
Eppendorf
®
Twin tec PCR plates, 96 wells,
semi-skirted, with barcode, def. color (wells colorless), pkg of 25 ea
Expand
View Pricing
EP0030128680
Eppendorf
®
Twin tec PCR plates, 96 wells,
red plate, conical bottom red wells, skirted, pkg of 25 ea
Expand
View Pricing
EP0030128842
Eppendorf
®
Twin tec PCR plates, 96 wells,
blue plate, conical bottom colorless wells, skirted, pkg of 300 ea
Expand
View Pricing
EP0030128656
Eppendorf
®
Twin tec PCR plates, 96 wells,
yellow plate, conical bottom colorless wells, skirted, pkg of 25 ea
Expand
View Pricing
EP0030128877
Eppendorf
®
Twin tec PCR plates, 96 wells,
semi-skirted, with barcode, def. color (wells colorless), pkg of 300 ea
Expand
View Pricing
EP0030128796
Eppendorf
®
Twin tec PCR plates, 96 wells,
green plate, conical bottom colorless wells, pkg of 300 ea
Expand
View Pricing
EP0030128583
Eppendorf
®
Twin tec PCR plates, 96 wells,
yellow plate, conical bottom yellow wells, semi-skirted, pkg of 25 ea
Expand
View Pricing
EP0030128990
Eppendorf
®
Twin tec PCR plates, 96 wells,
yellow plate, conical bottom colorless wells, skirted, pkg of 300 ea
Expand
View Pricing
EP0030128788
Eppendorf
®
Twin tec PCR plates, 96 wells,
yellow plate, conical bottom colorless wells, skirted, pkg of 300 ea
Expand
View Pricing
EP0030128664
Eppendorf
®
Twin tec PCR plates, 96 wells,
green plate, conical bottom colorless wells, skirted, pkg of 25 ea
Expand
View Pricing
EP0030128460
Eppendorf
®
Twin tec PCR plates, 96 wells,
skirted, with barcode, individual color (wells colorless), pkg of 25 ea
Expand
View Pricing
EP0030128850
Eppendorf
®
Twin tec PCR plates, 96 wells,
red plate, conical bottom colorless wells, skirted, pkg of 300 ea
Expand
View Pricing