Key Documents
EP0030128605
Eppendorf® Twin tec PCR plates, 96 wells
blue plate, conical bottom colorless wells, semi-skirted, pkg of 25 ea
About This Item
UNSPSC Code:
41106300
NACRES:
NB.15
material
blue plate
conical bottom colorless wells
polycarbonate
sterility
non-sterile
feature
skirt (Semi skirted)
packaging
pkg of 25 ea
manufacturer/tradename
Eppendorf® 0030128605
technique(s)
PCR: suitable
size
96 wells
well maximum volume
250 μm
working volume
250 μL
General description
  • Fits most thermal cyclers, 250 μL max. well volume (when used with cap strips).
  • Improved well-to-well tolerance.
  • Available with barcode (upon request), for skirted and semi-skirted only.
  • Ideal for quantitative real-time PCR as well − OptiTrack® matrix for faster sample identification and fewer pipetting errors.
Choose Eppendorf twin.tec PCR plates for confidence in every PCR assay. twin.tec plates offer purity, rigidity, color, and other simple, smart features can make your daily life a lot easier. The rigid polycarbonate frame works well in automation and high-throughput processes, and the different colors mean you can quickly organize the different steps of your workflow.
Features and Benefits
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Improved well-to-well tolerance, Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
  • Certified free of any detectable human DNA, DNase, RNase and PCR inhibitors).
  • Ideal for quantitative real-time PCR as well.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG