Key Documents
EP0030128680
Eppendorf® Twin tec PCR plates, 96 wells
red plate, conical bottom red wells, skirted, pkg of 25 ea
About This Item
UNSPSC Code:
41106300
material
conical bottom red wells
polycarbonate
red plate
sterility
non-sterile
feature
skirt (Skirted)
packaging
pkg of 25 ea
manufacturer/tradename
Eppendorf® 0030128680
technique(s)
PCR: suitable
size
96 wells
well maximum volume
150 μm
General description
Choose Eppendorf twin.tec PCR plates for confidence in every PCR assay. twin.tec plates offer purity, rigidity, color, and other simple, smart features can make your daily life a lot easier. The rigid polycarbonate frame works well in automation and high-throughput processes, and the different colors mean you can quickly organize the different steps of your workflow.
Application
Eppendorf® Twin tec PCR plates, 96 wells are ideal for quantitative real-time PCR.
Features and Benefits
  • A combination of polypropylene wells and torque-resistant polycarbonate frame
  • DNA, DNAse, RNAse, and PCR inhibitors free
  • Extremely thin-walled polypropylene wells ensure optimal heat transfer to the samples
  • Alphanumeric grid-referencing and cut-away corner
  • Effective sealing and reduced risk of contamination due to raised well rims
  • Easy stacking
  • Upon request available with a barcode
  • Low-profile design enables high efficiency for small sample volumes and enhances PCR efficiency
  • Eight holes in the skirt aid plate and easy removal from the thermal cycler block
  • Compatible with automated systems
  • Support a snug fit to the PCR cycler and act as a solid base for tight sealing
Legal Information
Eppendorf is a registered trademark of Eppendorf AG