Skip to Content
Merck

Skip To

1609002

USP

Salicylic acid

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-Hydroxybenzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
2-(HO)C6H4CO2H
CAS Number:
Molecular Weight:
138.12
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
774890
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

vapor density

4.8 (vs air)

vapor pressure

1 mmHg ( 114 °C)

API family

salicylic acid

manufacturer/tradename

USP

bp

211 °C (lit.)

mp

158-161 °C (lit.)

solubility

alcohol: freely soluble, benzene: slightly soluble, boiling water: soluble, chloroform: sparingly soluble, ether: freely soluble, toluene: slightly soluble, water: slightly soluble

density

1.44 g/cm3 at 20 °C

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC(=O)c1ccccc1O

InChI

1S/C7H6O3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H,(H,9,10)

InChI key

YGSDEFSMJLZEOE-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Salicylic acid USP reference standard for use in specified quality tests and assays.

Also used to prepare standard solutions for the assay, impurity analysis, and performance test by using liquid chromatography in conjunction with a UV detector according to the given below monographs of United States Pharmacopeia (USP) :
  • Salicylic Acid
  • Methyl Salicylate
  • Magnesium Salicylate Tablets
  • Aspirin Boluses

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S0200000PHR10131.00631
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

-

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

-

format

neat

format

neat

format

neat

format

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

liquid formulation
pharmaceutical

vapor density

4.8 (vs air)

vapor density

4.8 (vs air)

vapor density

4.8 (vs air)

vapor density

-

API family

salicylic acid

API family

salicylic acid

API family

salicylic acid

API family

-


Still not finding the right product?

Explore all of our products under Salicylic acid


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class

11 - Combustible Solids

flash_point_f

314.6 °F - closed cup

flash_point_c

157 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Methyl Salicylate
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 46(3) (2021)
Salicylic Acid
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 39(6), 3970-3970 (2020)
Magnesium Salicylate Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(4), 2724-2724 (2015)



Global Trade Item Number

SKUGTIN
1609002-125MG04061838745781

Questions

Reviews

No rating value

Active Filters