Skip to Content
Merck
All Photos(1)

Documents

PHR1013

Supelco

Salicylic acid

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2-Hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2H
CAS Number:
Molecular Weight:
138.12
Beilstein:
774890
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 775
traceable to Ph. Eur. S020000
traceable to USP 1609002

vapor density

4.8 (vs air)

vapor pressure

1 mmHg ( 114 °C)

API family

salicylic acid

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

211 °C (lit.)

mp

158-161 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

OC(=O)c1ccccc1O

InChI

1S/C7H6O3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H,(H,9,10)

InChI key

YGSDEFSMJLZEOE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Salicylic acid is an important material in pharmaceutical industry, obtained via the decomposition of acetylsalicylic acid. Salicylic acid is widely used in food industry for producing dyes and artificial scents. It also shows therapeutic potential in protecting the skin from sun damage and hence finds applications in cosmetics and medicine.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Salicylic acid may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using spectrofluorometric technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3036 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nickel?aluminum layered double hydroxide as a nanosorbent for selective solid-phase extraction and spectrofluorometric determination of salicylic acid in pharmaceutical and biological samples
Abdolmohammad-Zadeh H, et al.
Talanta, 84(2), 368-373 (2011)
Simultaneous determination of naproxen, salicylic acid and acetylsalicylic acid by spectrofluorimetry using partial least-squares (PLS) multivariate calibration
Navalon A, et al.
Talanta, 48(2), 469-475 (1999)
Shifeng Cao et al.
Food chemistry, 134(4), 1715-1718 (2013-02-28)
The effect of elicitors associated with host defence on betacyanin accumulation in Amaranthus mangostanus seedlings was investigated. Under the conditions of the experiments, betacyanin accumulation was generally enhanced by light. Methyl jasmonate (MeJA) treatment increased betacyanin synthesis in a concentration-dependent
Ganesan Karthikeyan et al.
British journal of haematology, 146(2), 142-149 (2009-05-15)
Guidelines differ on whether acetyl salicylic acid (ASA, aspirin) should be used for prophylaxis in patients at high-risk of venous thromboembolism (VTE), principally because of differences in perceptions of its efficacy. ASA is an attractive therapeutic option because it is
A Corina Vlot et al.
Annual review of phytopathology, 47, 177-206 (2009-04-30)
For more than 200 years, the plant hormone salicylic acid (SA) has been studied for its medicinal use in humans. However, its extensive signaling role in plants, particularly in defense against pathogens, has only become evident during the past 20

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service