Skip to Content
Merck
All Photos(1)

Key Documents

1255001

USP

Estrone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Estrone, 1,3,5(10)-Estratrien-3-ol-17-one, 3-Hydroxy-1,3,5(10)-estratrien-17-one, Folliculin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22O2
CAS Number:
Molecular Weight:
270.37
Beilstein:
1915077
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

estrone

manufacturer/tradename

USP

mp

258-260 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CCC2=O

InChI

1S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-16,19H,2,4,6-9H2,1H3/t14-,15-,16+,18+/m1/s1

InChI key

DNXHEGUUPJUMQT-CBZIJGRNSA-N

Gene Information

human ... ESR1(2099)

Looking for similar products? Visit Product Comparison Guide

Application

Estrone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Conjugated Estrogens
  • Conjugated Estrogens Tablets
  • Esterified Estrogens
  • Esterified Estrogens Tablets
  • Estradiol
  • Estradiol and Norethindrone Acetate Tablets
  • Estradiol Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert E Nelson et al.
Clinical chemistry, 50(2), 373-384 (2003-12-06)
Estradiol (E2) and estrone (E1) measurements form an integral part of the assessment of female reproductive function and have expanding roles in other fields. However, many E1 and E2 immunoassays have limited functional sensitivity, suffer from cross-reactivity, and display poor
Fluorometric determination of plasma 11-hydroxycorticosteroids. II. Studies on the specificity of the method.
L E Mejer et al.
Clinical chemistry, 19(7), 718-724 (1973-07-01)
Eva Fetter et al.
Aquatic toxicology (Amsterdam, Netherlands), 154, 221-229 (2014-06-14)
Xenoestrogens may persist in the environment by binding to sediments or suspended particulate matter serving as long-term reservoir and source of exposure, particularly for organisms living in or in contact with sediments. In this study, we present for the first
Xavier Remesar et al.
Medicinal research reviews, 32(6), 1263-1291 (2011-02-03)
Oleoyl-estrone (OE) is a powerful slimming agent that is also present in plasma and adipose tissue, where it is synthesized. It acts through the formation of a derivative W. OE effects (and W levels) are proportional to the dose. OE
Shanshan Zhao et al.
Breast cancer research : BCR, 16(2), R30-R30 (2014-03-29)
Paradoxically, a breast cancer risk reduction with conjugated equine estrogens (CEE) and a risk elevation with CEE plus medroxyprogesterone acetate (CEE + MPA) were observed in the Women's Health Initiative (WHI) randomized controlled trials. The effects of hormone therapy on serum sex

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service