Skip to Content
Merck
All Photos(1)

Documents

1142107

USP

Clozapine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

8-Chloro-11-(4-methyl-1-piperazinyl)-5H-dibenzo[b,e][1,4]-diazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19ClN4
CAS Number:
Molecular Weight:
326.82
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

clozapine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CN1CCN(CC1)C2=Nc3cc(Cl)ccc3Nc4ccccc24

InChI

1S/C18H19ClN4/c1-22-8-10-23(11-9-22)18-14-4-2-3-5-15(14)20-16-7-6-13(19)12-17(16)21-18/h2-7,12,20H,8-11H2,1H3

InChI key

QZUDBNBUXVUHMW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clozapine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Clozapine Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raúl Rigo-Bonnin et al.
Biochemia medica, 31(1), 010501-010501 (2021-01-01)
Clinicians request a large part of measurements of biological quantities that clinical laboratories perform for diagnostic, prognostic or diseases monitoring purposes. Thus, laboratories need to provide patient's results as reliable as possible. Metrological concepts like measurement uncertainty and metrological traceability
Colin M Cleary et al.
eLife, 10 (2021-05-21)
Glutamatergic neurons in the retrotrapezoid nucleus (RTN) function as respiratory chemoreceptors by regulating breathing in response to tissue CO2/H+. The RTN and greater parafacial region may also function as a chemosensing network composed of CO2/H+-sensitive excitatory and inhibitory synaptic interactions.
Benjamin W J Spencer et al.
Pharmacogenomics, 14(15), 1907-1914 (2013-11-19)
Clozapine is a uniquely efficacious antipsychotic drug in treatment-resistant schizophrenia. Its use is restricted due to adverse effects including a rare but dangerous reduction in neutrophils (agranulocytosis) and the mandatory hematological monitoring this entails in many countries. We review the
A Merenlender-Wagner et al.
Molecular psychiatry, 20(1), 126-132 (2013-12-25)
Autophagy is a process preserving the balance between synthesis, degradation and recycling of cellular components and is therefore essential for neuronal survival and function. Several key proteins govern the autophagy pathway including beclin1 and microtubule associated protein 1 light chain
Saeko Ikai et al.
The Annals of pharmacotherapy, 47(7-8), e31-e31 (2013-06-13)
To report on a patient who was successfully rechallenged with clozapine after perforation of the large intestine and pulmonary embolism post operatively, and provide a literature review on clozapine rechallenge. A 46-year-old Japanese man with treatment-resistant schizophrenia developed constipation and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service