Merck
  • Opening Pandora's box in the UK: a hypothetical pharmacogenetic test for clozapine.
Opening Pandora's box in the UK: a hypothetical pharmacogenetic test for clozapine.
Pharmacogenomics (2013-11-19)
Benjamin W J Spencer, Barbara Prainsack, Dan Rujescu, Ina Giegling, David A Collier, Fiona Gaughran, James H MacCabe, Cathy L Barr, Engilbert Sigurdsson, Henrik Stovring, Anil K Malhotra, Sarah R Curran
ABSTRACT
Clozapine is a uniquely efficacious antipsychotic drug in treatment-resistant schizophrenia. Its use is restricted due to adverse effects including a rare but dangerous reduction in neutrophils (agranulocytosis) and the mandatory hematological monitoring this entails in many countries. We review the statistical, ethical and legal issues arising from a hypothetical pharmacogenetic test for clozapine, using the UK as an exemplary case for consideration. Our key findings include: a consideration of the probabilistic results that a pharmacogenetic test may return; the impact on drug licensing; and the potential for pharmacogenetic tests for clozapine being used without consent under the UK's legal framework. We make recommendations regarding regulatory changes applicable to the special case of pharmacogenetic testing in clozapine treatment.
Product Description
Clozapine for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Clozapine Resolution Mixture, United States Pharmacopeia (USP) Reference Standard
Supelco
Clozapine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Clozapine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Clozapine
USP
Clozapine, United States Pharmacopeia (USP) Reference Standard