G004504
HPLC Analysis of Methylphenidate (Ritalin) Enantiomers on Astec® CHIROBIOTIC® V2
application for HPLC
About This Item
technique(s)
HPLC: suitable
test parameters
column: CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 μm particles (15024AST)
mobile phase: 15 mM ammonium acetate in methanol
flow rate: 1 mL/min
column temp.: 25 °C
detector: UV, 215 nm
injection: 5 μL
sample: 1 mg/mL in methanol
suitability
application for HPLC
application(s)
forensics and toxicology
pharmaceutical (small molecule)
Legal Information
Analyte
analytical column
standard
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service