Skip to Content
Merck
All Photos(2)

Key Documents

M2892

Sigma-Aldrich

Methylphenidate hydrochloride

Synonym(s):

threo-Methyl α-phenyl-α-(2-piperidyl)acetate hydrochloride, Ritalin hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19NO2 · HCl
CAS Number:
Molecular Weight:
269.77
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77

drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

application(s)

forensics and toxicology

originator

Johnson & Johnson

SMILES string

Cl[H].COC(=O)C(C1CCCCN1)c2ccccc2

InChI

1S/C14H19NO2.ClH/c1-17-14(16)13(11-7-3-2-4-8-11)12-9-5-6-10-15-12;/h2-4,7-8,12-13,15H,5-6,9-10H2,1H3;1H

InChI key

JUMYIBMBTDDLNG-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

CNS stimulant; releases catecholamines; potent inhibitor of dopamine reuptake

Features and Benefits

This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert Lew et al.
Psychopharmacology, 237(11), 3435-3446 (2020-08-20)
Drugs that rapidly increase dopamine levels have an increased risk of abuse. Dasotraline (DAS) is a dopamine and norepinephrine reuptake inhibitor characterized by slow oral absorption with low potential for abuse. However, it remains unclear whether intravenous (i.v.) administration would
R Kuczenski et al.
The Journal of pharmacology and experimental therapeutics, 296(3), 876-883 (2001-02-22)
The prescribed use of methylphenidate (Ritalin) in the treatment of attention deficit hyperactivity disorder has risen dramatically in recent years. The relative roles of dopamine, norepinephrine, and serotonin in the therapeutic action of these drugs was assessed by comparing the
Eduardo Bruera et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(19), 2421-2427 (2013-05-22)
Cancer-related-fatigue (CRF) is common in advanced cancer. The primary objective of the study was to compare the effects of methylphenidate (MP) with those of placebo (PL) on CRF as measured using the Functional Assessment of Chronic Illness Therapy-Fatigue (FACIT-F) fatigue
Kleber Santos et al.
Developmental medicine and child neurology, 55(7), 654-660 (2013-03-14)
Difficult-to-treat epilepsies and attention-deficit-hyperactivity disorder (ADHD) often co-occur. Because of concerns about the use of stimulants in children with this comorbidity, the impact of ADHD treatment on seizure frequency and severity is not known. This pilot study evaluated the safety
Natalie Grizenko et al.
Canadian journal of psychiatry. Revue canadienne de psychiatrie, 58(11), 632-639 (2013-11-20)
Examine the short-term (acute) effects of methylphenidate (MPH) on math performance in children with attention-deficit hyperactivity disorder (ADHD) and what factors predict improvement in math performance. One hundred ninety-eight children with ADHD participated in a double-blind, placebo-controlled, randomized crossover MPH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service