Skip to Content
Merck
All Photos(1)

Documents

57317

Supelco

SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB)

df 60 μm(PDMS/DVB, for use with autosampler/HPLC, needle size 24 ga, StableFlex fiber

Synonym(s):

SPME/HPLC FIBER PK3 W/60UM PDMS-DVB COATING

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

material

StableFlex fiber
notched brown hub

needle size

24 ga

packaging

pkg of 3 ea

df

60 μm (PDMS/DVB)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

1 cm

matrix active group

PDMS/DVB coating

application(s)

food and beverages

compatibility

for analyte group volatiles, amines, and nitroaromatic compounds (MW 50-300)
for use with autosampler/HPLC

Looking for similar products? Visit Product Comparison Guide

Application

Amines and Polar Compounds

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pawliszyn, Janusz
Solid Phase Microextraction: Theory and Practice, 264-264 (1997)
Pawliszyn, Janusz
Handbook of Solid Phase Microextraction, 410-410 (2009)
Pawliszyn, Janusz
Applications of Solid Phase Microextraction, 674-674 (1999)
Talon, R., Montel, M.
Applied SPME, 364-371 (1999)
Edited by Sue Ann Scheppers Wercinski
Solid Phase Microextraction: A Practical Guide, 257-257 (1999)

Related Content

A demonstration of the use of innovative BioSPME fibers for the selective extraction of drugs and polar metbolites from biological matrices.

Chromatograms

application for SPME, application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service