Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB)
Products
Brand
Supelco (2)
Sustainability
Feature
Length (cm)
Application
Color
d
f
(µm)
Gauge
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
57311
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB),
d
f
65 μm(PDMS/DVB, needle size 24 ga, for use with autosampler
Expand
View Pricing
57902-U
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB),
d
f
65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga, metal alloy fiber
Expand
View Pricing