Skip to Content
Merck
All Photos(1)

Documents

46323-U

Supelco

Aflatoxin B1 solution

3 μg/mL in benzene:acetonitrile (98:2), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12O6
CAS Number:
Molecular Weight:
312.27
Beilstein:
1269174
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 1 mL

concentration

3 μg/mL in benzene:acetonitrile (98:2)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

[H][C@]12OC=C[C@@]1([H])c3c(O2)cc(OC)c4C5=C(C(=O)CC5)C(=O)Oc34

InChI

1S/C17H12O6/c1-20-10-6-11-14(8-4-5-21-17(8)22-11)15-13(10)7-2-3-9(18)12(7)16(19)23-15/h4-6,8,17H,2-3H2,1H3/t8-,17+/m0/s1

InChI key

OQIQSTLJSLGHID-WNWIJWBNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aflatoxin B1 (AFB1) is a carcinogenic compound that induces transversion of G to T at codon 249 of the p53 tumor suppressor gene. AFB1 is the most common in food and amongst the most potent genotoxic and carcinogenic aflatoxins. It is produced by both Aspergillus flavus and A. parasiticus.

Application

AFB1 solution was used as reference standard for the analysis of mycotoxins in paprika using LC-FD and LC-MS. AFB1 may be used as spiking standard solution to compare and determine AFB1 in corn and peanut samples using ultra-high-pressure liquid chromatography with UV detection.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Zhaohui Fu et al.
Journal of chromatography. A, 1209(1-2), 271-274 (2008-10-07)
A rapid and simple method using ultra-high-pressure liquid chromatography with UV detection for the determination of aflatoxins B1, B2, G1 and G2 in corn and peanuts has been developed. In this method, aflatoxins were extracted with a mixture of acetonitrile
F Aguilar et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(18), 8586-8590 (1993-09-15)
Approximately half of hepatocellular carcinoma (HCC) from regions in the world with high contamination of food with the mycotoxin aflatoxin B1 (AFB1) contain a mutation in codon 249 of the p53 tumor suppressor gene. The mutation almost exclusively consists of
J Miguel Hernández Hierro et al.
Journal of agricultural and food chemistry, 56(3), 751-756 (2008-01-22)
Aflatoxins are the only mycotoxins with legal limits for spices in the European Union. A further limit for ochratoxin A is expected to be adopted soon. Thus, rapid simultaneous methods for quantifying the five mycotoxins are sought. Liquid extraction, immunoaffinity
R L Cravens et al.
Poultry science, 92(8), 1997-2004 (2013-07-23)
The effects of increasing aflatoxin B1 concentration (0, 0.75, 1.5 mg/kg) on broilers with or without necrotic enteritis or virginiamycin were determined. In the 23-d study, 22 male Cobb 500 chicks per pen were allotted to 12 treatments (3 ×
D V Neeff et al.
Poultry science, 92(1), 131-137 (2012-12-18)
The aim of this study was to determine the binding capacity of a hydrated sodium calcium aluminosilicate (HSCAS) for aflatoxin B(1) (AFB(1)), and the efficacy of the HSCAS to reduce the concentrations of residual AFB(1) and its metabolites in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service