Skip to Content
Merck
All Photos(1)

Key Documents

34034

Supelco

Aflatoxin B2 solution

0.5 μg/mL in acetonitrile, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14O6
CAS Number:
Molecular Weight:
314.29
Beilstein:
1355115
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

0.5 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

[H][C@@]12CCO[C@]1([H])Oc3cc(OC)c4C5=C(C(=O)CC5)C(=O)Oc4c23

InChI

1S/C17H14O6/c1-20-10-6-11-14(8-4-5-21-17(8)22-11)15-13(10)7-2-3-9(18)12(7)16(19)23-15/h6,8,17H,2-5H2,1H3/t8-,17+/m0/s1

InChI key

WWSYXEZEXMQWHT-WNWIJWBNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aflatoxin B2 is one of the fungal metabolic products produced in food and feed, contaminated by Aspergillus species.
Certain vial.
Aflatoxins are mycotoxins that are generated as a result of secondary metabolism of various fungal species such as Aspergillus, Penicillium and Fusarium.
Certan Vial

Application

Aflatoxin B2 may be used for the determination of aflatoxin B2 in:
  • Animal feed by high performance liquid chromatography with fluorescence detection (HPLC-FLD) as well as quick, easy, cheap, effective, rugged and safe (QuEchERS) extraction method combined with ultra-high performance liquid chromatography-high resolution mass spectrometry (UHPLC-HRMS).
  • Eggs by QuEchERS extraction method followed by analysis using UHPLC with tandem mass spectrometry (MS/MS) operating on electrospray-ionization interface (ESI) mode.
  • Brazil nuts by immunoaffinity column (IAC) and clean-up HPLC-FLD.
  • Plant-based milk and dairy products by dispersive liquid-liquid microextraction (DLLME), QuEchERS extraction method and HPLC-FLD.

Aflatoxin B2 solution may be used as an analytical reference standard for the determination of the analyte in meat, meat products, eggs and milk using high-performance liquid chromatography (HPLC) with fluorescence and ultraviolet detection.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Debora Muratori Holanda et al.
Toxins, 12(8) (2020-08-13)
Pigs are highly susceptible to mycotoxins. This study investigated the effects of a postbiotic yeast cell wall-based blend (PYCW; Nicholasville, KY, USA) on growth and health of newly-weaned pigs under dietary challenge of multiple mycotoxins. Forty-eight newly-weaned pigs (21 d
Determination of aflatoxins in eggs, milk, meat and meat products using HPLC fluorescent and UV detectors.
Herzallah MS.
Food Chemistry, 114(3), 1141-1146 (2009)
Extended validation of a sensitive and robust method for simultaneous quantification of aflatoxins B1, B2, G1 and G2 in Brazil nuts by HPLC-FLD.
Campos WEO, et al.
J. Food Compos. Anal., 60(10), 90-96 (2017)
Target analysis and retrospective screening of veterinary drugs, ergot alkaloids, plant toxins and other undesirable substances in feed using liquid chromatography-high resolution mass spectrometry.
Leon N, et al.
Talanta, 149(1), 43-52 (2016)
Ultra-high performance liquid chromatography tandem mass spectrometry for simultaneous analysis of aflatoxins B1, G1, B2, G2, zearalenone and its metabolites in eggs using a QuEChERS-based extraction procedure.
Li Y, et al.
Analytical Methods : Advancing Methods and Applications, 7(10), 4145-4151 (2015)

Protocols

LC/MS/MS Analysis of Aflatoxins in Hops on Ascentis® Express 2.7 μm Phenyl-Hexyl after Cleanup Using Supel™ Tox AflaZea SPE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service