Skip to Content
Merck
All Photos(5)

Key Documents

T9900

Sigma-Aldrich

Methyl tricosanoate

≥99.0% (GC)

Synonym(s):

Tricosanoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)21COOCH3
CAS Number:
Molecular Weight:
368.64
Beilstein:
1796627
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

synthetic (organic)

Quality Level

Assay

≥99.0% (GC)

form

powder

technique(s)

gas chromatography (GC): suitable

color

white

mp

53-56 °C (lit.)

solubility

chloroform: 50 mg/mL, clear, colorless

application(s)

agriculture
general analytical

functional group

ester

lipid type

saturated FAs

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C24H48O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24(25)26-2/h3-23H2,1-2H3

InChI key

VORKGRIRMPBCCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl tricosanoate, also recognized as Tricosanoic acid methyl ester, represents the fatty acid methyl ester (FAME) form of tricosanoic acid. This odd-numbered long-chain fatty acid methyl ester is atypical in many biological systems, making it valuable as a biomarker. Tricosanoic acid is naturally synthesized in certain plants and mammals, including humans, where it predominates as a skin lipid. It can be found acylated to various glyco-, phospho-, and sphingolipids, including gangliosides. This fatty acid holds significance in biochemical, lipidomics, and metabolomics research applications.

Application

Methyl tricosanoate has been used as an internal standard to quantify fatty acids or fatty acid methyl esters (FAMEs).

Features and Benefits

  • High-quality molecule suitable for mulitple research applications
  • Commonly employed in Metabolomics and Biochemical studies

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carolina Oliveira de Souza et al.
Lipids in health and disease, 16(1), 78-78 (2017-04-16)
Palm olein is used in infant formula fat blends in order to match the fatty acid profile of human milk. While the effects on fatty acid balance have been evaluated, the use of palm olein in combination with palm kernel
Daniel L Forwood et al.
Meat science, 173, 108402-108402 (2020-12-15)
This study investigated the effect of feeding unsaleable carrots to lambs within a total-mixed ration (TMR) on performance, carcass characteristics, meat quality and sensory parameters. Thirty-six Australian Merino wether lambs were fed a control (barley-based) or carrot-based TMR for 11-weeks.
K E Aarak et al.
Food & function, 5(9), 2331-2337 (2014-08-01)
The aim of the present work was to study the effect of a broccoli phytochemical extract (Br-ex) on the release of fatty acids (FA) from salmon muscle (SM) and salmon oil (SO) during in vitro digestion. The hypothesis of the
Marine Vallet et al.
Marine drugs, 18(8) (2020-07-28)
Kelps are colonized by a wide range of microbial symbionts. Among them, endophytic fungi remain poorly studied, but recent studies evidenced yet their high diversity and their central role in algal defense against various pathogens. Thus, studying the metabolic expressions
Lihong Zhou et al.
Journal of proteome research, 14(5), 2331-2347 (2015-04-01)
Silkworm (Bombyx mori) is a lepidopteran-holometabolic model organism. To understand its developmental biochemistry, we characterized the larval hemolymph metabonome from the third instar to prepupa stage using (1)H NMR spectroscopy whilst hemolymph fatty acid composition using GC-FID/MS. We unambiguously assigned

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service