Skip to Content
Merck
All Photos(1)

Key Documents

T0627

Sigma-Aldrich

Methyl tridecanoate

≥97% (GC)

Synonym(s):

Tridecanoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11COOCH3
CAS Number:
Molecular Weight:
228.37
Beilstein:
1769695
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (GC)

form

liquid

refractive index

n20/D 1.435 (lit.)

bp

131 °C/4 mmHg (lit.)

mp

5.5 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

functional group

ester

lipid type

saturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC(=O)OC

InChI

1S/C14H28O2/c1-3-4-5-6-7-8-9-10-11-12-13-14(15)16-2/h3-13H2,1-2H3

InChI key

JNDDPBOKWCBQSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K J Phelps et al.
Journal of animal science, 93(3), 1298-1308 (2015-05-29)
The objective of this study was to examine the effects of alternative finishing strategies on feedlot performance and carcass characteristics. Beef steers (64 pens; 8 steers/pen) were allocated to a randomized complete block design with a 2 × 2 factorial
Joonhoon Kim et al.
Frontiers in bioengineering and biotechnology, 8, 612832-612832 (2021-02-16)
An oleaginous yeast Rhodosporidium toruloides is a promising host for converting lignocellulosic biomass to bioproducts and biofuels. In this work, we performed multi-omics analysis of lignocellulosic carbon utilization in R. toruloides and reconstructed the genome-scale metabolic network of R. toruloides.
C B Paulk et al.
Journal of animal science, 93(4), 1666-1678 (2015-05-29)
Data from existing literature were used to generate equations to predict finishing pig back, belly, and jowl fat iodine values (IV) and an experiment was conducted to evaluate these equations. The final database included 24, 21, and 29 papers for
Maria Teresa Osorio et al.
Food chemistry, 189, 67-73 (2015-07-21)
European Regulation 1169/2011 requires producers of foods that contain refined vegetable oils to label the oil types. A novel rapid and staged methodology has been developed for the first time to identify common oil species in oil blends. The qualitative
C A Alvarado-Gilis et al.
Journal of animal science, 93(9), 4368-4376 (2015-10-07)
The objective of this study was to evaluate the efficacy of protecting PUFA within ground flaxseed against ruminal biohydrogenation by encapsulating them in a matrix consisting of a 1:1 blend of ground flaxseed and dolomitic lime hydrate (L-Flaxseed). Crossbreed heifers

Protocols

Protocol for GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service