Skip to Content
Merck
All Photos(1)

Documents

T8019

Sigma-Aldrich

Tentoxin from Alternaria tenuis

Naturally occurring phytotoxic cyclic tetrapeptide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30N4O4
CAS Number:
Molecular Weight:
414.50
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.56

biological source

fungus (Alternaria tenuis)

Quality Level

Assay

≥95% (HPLC)

storage temp.

2-8°C

SMILES string

CC(C)CC1NC(=O)C(C)N(C)C(=O)CNC(=O)\C(=C/c2ccccc2)N(C)C1=O

InChI

1S/C22H30N4O4/c1-14(2)11-17-22(30)26(5)18(12-16-9-7-6-8-10-16)21(29)23-13-19(27)25(4)15(3)20(28)24-17/h6-10,12,14-15,17H,11,13H2,1-5H3,(H,23,29)(H,24,28)/b18-12-/t15-,17-/m0/s1

InChI key

SIIRBDOFKDACOK-LFXZBHHUSA-N

General description

Tentoxin is a majorly occurring cyclic tetrapeptide mycotoxin that is excreted by the Alternaria fungal species like Alternaria alternata and Alternaria tenuis.

Biochem/physiol Actions

It induces chlorosis in germinating seedlings of many dicotyledonous plants. Tentoxin has been postulated to inhibit cyclic photophosphorylation by acting as an energy transfer inhibitor at the terminal steps of ATP synthesis and to target the F1 moiety of photosynthetic H+-ATPases. A kinetic analysis of the action of tentoxin on the chloroplast F1 H+-ATPase (CF1) portion of chloroplast ATP synthase has been reported.
Tentoxin is phytotoxic, which induces chlorosis in plant leaves and germinating seedlings. It also halts photophosphorylation by inhibiting chloroplastic ATP synthase. Tentoxin over energizes thylakoids and is converted to isotentoxin by UV irradiation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Sigalat et al.
FEBS letters, 368(2), 253-256 (1995-07-17)
The effect of tentoxin at high concentrations was investigated in thylakoids and proteoliposomes containing bacteriorhodopsin and CF0CF1. Venturicidin-sensitive ATP hydrolysis, ATP-generated delta pH and ATP synthesis were practically 100% inhibited at 2 microM tentoxin, and restored to various extents beyond
Inhibition of photophosphorylation by tentoxin, a cyclic tetrapeptide.
C J Arntzen
Biochimica et biophysica acta, 283(3), 539-542 (1972-12-14)
Jingfeng Lou et al.
Molecules (Basel, Switzerland), 18(5), 5891-5935 (2013-05-24)
Alternaria is a cosmopolitan fungal genus widely distributing in soil and organic matter. It includes saprophytic, endophytic and pathogenic species. At least 268 metabolites from Alternaria fungi have been reported in the past few decades. They mainly include nitrogen-containing metabolites
Cytotoxicity of Extracts from Alternaria alternata against Cultured Tobacco BY-2Cells
OH KB, et al.
Biocontrol Science, 7(2), 127-130 (2002)
Lionel Perrin et al.
The FEBS journal, 278(12), 2167-2178 (2011-04-27)
We report unambiguous proof of the stability of a carbinol intermediate in the case of P450 metabolism of an N-methylated natural cyclo-peptide, namely tentoxin. Under mild acidic or neutral conditions, the lifetime of carbinol-amide is long enough to be fully

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service