Skip to Content
Merck
All Photos(1)

Key Documents

T5639

Sigma-Aldrich

Ticarcillin disodium salt

Synonym(s):

(2S,5R,6R)-6-[[(2R)-Carboxy-3-thienylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14N2Na2O6S2
CAS Number:
Molecular Weight:
428.39
MDL number:
UNSPSC Code:
51282427
PubChem Substance ID:
NACRES:
NA.85
Pricing and availability is not currently available.

biological source

semisynthetic

Quality Level

form

powder or crystals

color

white to light yellow

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](C([O-])=O)c3ccsc3)C(=O)N2[C@H]1C([O-])=O

InChI

1S/C15H16N2O6S2.2Na/c1-15(2)9(14(22)23)17-11(19)8(12(17)25-15)16-10(18)7(13(20)21)6-3-4-24-5-6;;/h3-5,7-9,12H,1-2H3,(H,16,18)(H,20,21)(H,22,23);;/q;2*+1/p-2/t7-,8-,9+,12-;;/m1../s1

InChI key

ZBBCUBMBMZNEME-QBGWIPKPSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam

Application

Ticarcillin is a semisynthetic antibiotic with a broad spectrum of bactericidal activity against many gram-positive and gram-negative aerobic and anaerobic bacteria. It is commonly used in combination with clavulanate in order to improve it′s efficacy[1]. It is used to study appropriate dosing in pediatric patients who have cystic fibrosis[2].

Biochem/physiol Actions

Ticarcillin is a β-Lactam antibiotic that prevents bacterial cell wall biosynthesis at the level of peptidogylcan cross-linking by inhibiting peptidoglycan transpeptidases. It is derived from penicillin and is similar to carbenicillin in action. Ticarcillin is susceptible to degradation by ß-lactamases[1].

Packaging

1G

Other Notes

β-Lactam antibiotic
Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Keep in a dry place.

Storage and Stability

Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Keep in a dry place.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalie A Peartree et al.
Psychopharmacology, 234(12), 1815-1828 (2017-04-01)
Smoking typically begins during adolescence or early adulthood in a social context, yet the role of social context in animal models is poorly understood. The present study examined the effect of social context on acquisition of nicotine self-administration. Sixty-day-old male
R N Brogden et al.
Drugs, 20(5), 325-352 (1980-11-01)
Ticarcillin is a semisynthetic penicillin for parenteral administration. The antibacterial activity of ticarcillin is similar to that of carbenicillin except that it is two to four times more active in vitro against Pseudomonas aeruginosa, generally less active against Gram-positive cocci
Jennifer M Loftis et al.
European journal of pharmacology, 880, 173175-173175 (2020-05-18)
There are no medications that target the neurotoxic effects or reduce the use of methamphetamine. Recombinant T-cell receptor ligand (RTL) 1000 [a partial major histocompatibility complex (pMHC) class II construct with a tethered myelin peptide], addresses the neuroimmune effects of
Rocio Ochoa-Fernandez et al.
Nature methods, 17(7), 717-725 (2020-07-01)
Optogenetics is the genetic approach for controlling cellular processes with light. It provides spatiotemporal, quantitative and reversible control over biological signaling and metabolic processes, overcoming limitations of chemically inducible systems. However, optogenetics lags in plant research because ambient light required
Müserref Tatman-Otkun et al.
BMC microbiology, 5, 24-24 (2005-05-11)
Stenotrophomonas maltophilia is inherently resistant to many antimicrobials. So far, antimicrobial susceptibility tests for S. maltophilia have not been fully standardized. The purpose of the study was to compare the susceptibility of S. maltophilia isolates against seven different antimicrobials using

Questions

  1. Is the potency indicated for T5639 (CAS 4697-14-7) to be interpreted as the potency for the free form of Ticarcillin, or is it the potency of the salt form?

    1 answer
    1. The potency of T5639 is for the free form of Ticarcillin per USP.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service