SRP3314
IL-22 human
Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Synonym(s):
IL-TIF
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
biological source
human
recombinant
expressed in E. coli
Assay
≥98% (HPLC)
≥98% (SDS-PAGE)
form
lyophilized
potency
0.7-1.0 ng/mL
mol wt
33.6 kDa
packaging
pkg of 10 μg
impurities
endotoxin, tested
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... IL22(50616)
General description
IL-22 is a member of the IL-10 family of regulatory cytokines which includes IL-10, IL-19, IL-20, IL-22, IL-24 and IL-26. Members of this family share partial homology in their amino acid sequences, but they are dissimilar in their biological functions. Produced by T lymphocytes, IL-22 inhibits IL-4 production by Th2 cells, and induces acute phase reactants in the liver and pancreas. IL-22 signals through a receptor system consisting of IL-10R?/CRF2-4 and IL-22R, both of which are members of the class II cytokine-receptor family. Recombinant human IL-22 is a 33.6 kDa non-disulfide-linked homodimeric protein containing of two 147 amino acid polypeptide chains.
Biochem/physiol Actions
IL-22 is a member of the IL-10 family of regulatory cytokines which includes IL-10, IL-19, IL-20, IL-22, IL-24 and IL-26. Recombinant human IL-22 is a 33.6 kDa non-disulfide-linked homodimeric protein containing of two 147 amino acid polypeptide chains.
Physical form
Lyophilized from 10 mM NaCitrate, pH 4.0.
Reconstitution
Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.5-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Sergei V Kotenko
Cytokine & growth factor reviews, 13(3), 223-240 (2002-12-19)
Five novel cytokines (IL-19, IL-20, IL-22 (IL-TIF), IL-24 (human MDA-7, mouse FISP, rat C49A/Mob-5), and IL-26 (AK155)) demonstrating limited primary sequence identity and probable structural homology to IL-10 have been identified. These cellular cytokines, as well as several cytokines encoded
P Conti et al.
Immunology letters, 88(3), 171-174 (2003-08-28)
It has been reported that the CD4+ T cell is a very important source of interleukin 10 (IL-10), while CD8+ cells produce low amounts. IL-10 exerts several immune stimulating, as well as inhibitory effects. There are at least five novel
Yi Zhao et al.
International journal of clinical and experimental medicine, 8(6), 9580-9584 (2015-08-27)
The interleukin (IL)-23/IL-17A/IL-22 cytokine axis plays a critical role in the pathogenesis of psoriasis, wherein IL-22 effects on epidermal alternations by inhibiting differentiation and inducing keratinocyte proliferation. In this study, we investigated effects of curcumin on IL-22-induced proliferation in a
María J Loera-Arias et al.
Biotechnology letters, 36(12), 2489-2494 (2014-09-13)
Interleukin-22 (IL-22) participates in the modulation of innate immunity and inflammation. This cytokine has important therapeutic potential, such as with ulcerative colitis, liver and lung injury, and infection, in different animal models. We generated a Lactococcus lactis strain that secretes
H Fukui et al.
British journal of cancer, 111(4), 763-771 (2014-06-18)
Interleukin-22 (IL-22) has been recently highlighted owing to its biological significance in the modulation of tissue responses during inflammation. However, the role of IL-22 in carcinogenesis has remained unclear. Here, we investigated the pathophysiological significance of IL-22 expression in gastric
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service