Skip to Content
Merck
All Photos(1)

Key Documents

H7791

Sigma-Aldrich

IL-17A human

HumanKine®, recombinant, Xeno-free, >95% (SDS-PAGE), suitable for cell culture

Synonym(s):

CTLA-8, Interleukin-17A human, IL-17

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Product Name

Interleukin-17A human, IL-17A, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

≤2 ng/mL EC50

quality

endotoxin tested

mol wt

dimer 30-35 kDa (glycosylated)

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

≤1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL17A(3605)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

IL-17, initially known as CTLA-8, is a T cell-expressed polypeptide that appears to be involved in the regulation of the hematopoietic system. IL-17 has been shown to induce IL-6, IL-8, and G-CSF production by fibroblasts; all of these cytokines have demonstrated effects on hematopoiesis.
IL-17A belongs to the IL-17 family and is produced by Th17 cells, a subset of T helper cells. Interleukin 17 (IL-17), initially known as CTLA-8, is a T cell expressed polypeptide that appears to be involved in the regulation of the hematopoietic system. IL-17 has been shown to induce IL-6, IL-8, and G-CSF production by fibroblasts; all of these cytokines have demonstrated effects on hematopoiesis. IL-17 induces stromal cells to produce proinflammatory and hematopoietic cytokines, upregulates production of growth factors that promote angiogenesis, and enhances expression of adhesion molecules in fibroblasts.
Initially known as CTLA-8; is a T cell-expressed polypeptide that appears to be involved in the regulation of the hematopoietic system; has been shown to induce IL-6, IL-8, and G-CSF production by fibroblasts; all of these cytokines have demonstrated effects on hematopoiesis.

Physical form

Lyophilized from a 0.2 μm filtered solution of 10 mM Tris-HCl pH 7.4+ 250 mM NaCl

Preparation Note

HumanKine Interleukin-17A (IL-17A), expressed in human HEK 293 cells, is a glycosylated homodimer with an apparent molecular mass of 30-35 kDa due to glycosylation. Production in human 293 cells offers authentic glycosylation. Glycosylation contributes to stability in cell growth media and other applications.

Analysis Note

The specific activity was determined by the dose-dependent induction of IL-6 secretion from NDHF adult fibroblasts.

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shu Zhu et al.
The Journal of experimental medicine, 207(12), 2647-2662 (2010-11-17)
Interleukin 17 (IL-17) plays critical roles in the pathogenesis of various autoimmune diseases, including experimental autoimmune encephalomyelitis (EAE). How the signals triggered by this powerful inflammatory cytokine are controlled to avoid abnormal inflammatory responses is not well understood. In this
Stéphane Dragon et al.
American journal of respiratory cell and molecular biology, 50(6), 1053-1063 (2014-01-08)
Airway smooth muscle (ASM) cells are thought to contribute to the pathogenesis of allergic asthma by orchestrating and perpetuating airway inflammation and remodeling responses. In this study, we evaluated the IL-17RA signal transduction and gene expression profile in ASM cells
Michael Notaras et al.
Molecular psychiatry (2021-06-24)
It is widely accepted that narcotic use during pregnancy and specific environmental factors (e.g., maternal immune activation and chronic stress) may increase risk of neuropsychiatric illness in offspring. However, little progress has been made in defining human-specific in utero neurodevelopmental
Junhua Wang et al.
PLoS neglected tropical diseases, 9(5), e0003755-e0003755 (2015-05-09)
The growth potential of the tumor-like Echinococcus multilocularis metacestode (causing alveolar echinococcosis, AE) is directly linked to the nature/function of the periparasitic host immune-mediated processes. We previously showed that Fibrinogen-like-protein 2 (FGL2), a novel CD4+CD25+ Treg effector molecule, was over-expressed
Kelly M Adams et al.
PloS one, 9(8), e103263-e103263 (2014-08-21)
The epithelium plays an active role in the response to inhaled pathogens in part by responding to signals from the immune system. Epithelial responses may include changes in chemokine expression, increased mucin production and antimicrobial peptide secretion, and changes in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service