Skip to Content
Merck
All Photos(1)

Key Documents

SRP0373

Sigma-Aldrich

CDK4/Cyclin D1 active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥90% (SDS-PAGE)

Synonym(s):

BCL1, CCND1, Cell division protein kinase 4, Cyclin-dependent kinase 4, PRAD1, PSK-J3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

29 kDa (Cyclin D1)
34 kDa (CDK4)

packaging

pkg of 20 μg

solubility

water: soluble

application(s)

life science and biopharma

shipped in

dry ice

storage temp.

−70°C

Gene Information

General description

Research area: Apoptosis

Cyclin D1 (CCND1) is a proto-oncogene. It is one of the three D-type cyclins. The gene encoding it is localized on human chromosome 11q13. Cyclin dependent kinase-like 4 (CDKL4) is part of the cyclin-dependent protein kinase (CDK) family. The gene encoding it is localized on human chromosome 12q14-15.
Recombinant, full length human CDK4 (Genbank Accession No. NM_000075), amino acids 16-271, with C-terminal His-tag and modifications (amino acids 43G-47G were replaced with EE, and T172A), MW=34 kDa, and human Cyclin D1 (Genbank accession number NM_053056), MW 29 kDa, were co-expressed in a Baculovirus-infected Sf9 cells expression system.

Biochem/physiol Actions

Cyclin D1 (CCND1) regulates the Wnt signaling pathway. It also regulates the progression of the cell cycle from the G1 to S phase in many cell types. CCND1 phosphorylates and inactivates retinoblastoma (Rb) protein. Cyclin D1 has been associated with mantle cell lymphoma. This protein is overexpressed in multiple cancers like hepatocellular carcinoma. Cyclin dependent kinase-like 4 (CDKL4) has been associated with various tumors and colorectal cancer. The protein regulates apoptosis of nucleated cells by modifying transcriptional regulation of genes governing cell proliferation and cell death. Like CCND1, it also modulates the progression of cycle from G1 to S phase.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impact of cyclin-dependent kinase CDK4 inhibition on eryptosis.
Lang E
Cellular Physiology and Biochemistry, 37(3), 1178-1186 (2015)
Association Between Polymorphism rs678653 in Human Cyclin D1 Gene (CCND1) and Susceptibility to Cancer: A Meta-Analysis
Xichao Dai
Medical Science Monitor : International Medical Journal of Experimental and Clinical Research, 22, 863-874 (2016)
High-throughput RNAi screening of human kinases identifies predictors of clinical outcome in colorectal cancer patients treated with oxaliplatin
Moubin Lin
Oncotarget, 6(18), 16774-16785 (2015)
Bioinformatics prediction and experimental validation of microRNA-20a targeting Cyclin D1 in hepatocellular carcinoma
Hamzeh Karimkahnloo
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 39(4) (2017)
miR-16 induction after CDK4 knockdown is mediated by c-Myc suppression and inhibits cell growth as well as sensitizes nasopharyngeal carcinoma cells to chemotherapy.
Jiang Q
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 37(2), 2425-2433 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service