Skip to Content
Merck
All Photos(2)

Key Documents

RAB0311

Sigma-Aldrich

Rat IL-6 ELISA Kit

for serum, plasma and cell culture supernatants

Synonym(s):

B-cell stimulatory factor 2, CTL differentiation factor, Hybridoma growth factor, IFN-beta-2, IL-6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32
Pricing and availability is not currently available.

species reactivity

rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 30 pg/mL
standard curve range: 40.96-10000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... Il6(24498)

General description

This enzyme-linked immunosorbent assay (ELISA) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of a target protein in biological samples, such as serum, plasma, cell culture supernatants, urine and/or cell and tissue lysates.

Immunogen

Recombinant Rat IL-6

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Rat IL-6 ELISA Kit has been used to measure interleukin-6 (IL-6) levels in serum,[1] culture supernatants[2] and plasma.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protective Effects of Reduced Beta 2 Glycoprotein I on Liver Injury in Streptozotocin (STZ)-Diabetic Rats by Activation of AMP-Activated Protein Kinase
Zhang JY, et al.
Medical Science Monitor : International Medical Journal of Experimental and Clinical Research, 24, 7577-7577 (2018)
Maurilio da Silva Morrone et al.
Oxidative medicine and cellular longevity, 2016, 5719291-5719291 (2015-12-08)
The aim of this study was to investigate the potential of curcumin oral supplementation (50 and 100 mg/Kg/day, for 30 days) in circumventing menopause-associated oxidative stress and lipid profile dysfunctions in a rat ovariectomy (OVX) model. Female Wistar rats were operated
Ling Li et al.
Oncology letters, 15(2), 2550-2554 (2018-02-13)
The present study aimed to investigate the effect of tanshinone II, isolated from Salvia miltiorrhiza Bunge, on lipopolysaccharide (LPS)-induced acute lung injury (ALI) in rats. Male Sprague-Dawley rats were divided into three groups: Control, LPS and tanshinone II. Animals in
Zhiyan Tian et al.
Medical science monitor : international medical journal of experimental and clinical research, 24, 3922-3928 (2018-06-11)
BACKGROUND Ginsenoside is the major bioactive component of ginseng, which has been proven to be a neuroprotective drug. The aim of this study was to evaluate the therapeutic effect of ginsenoside in a diabetic Goto-Kakizaki (GK) rat model. MATERIAL AND
Yingjie Wang et al.
Molecular medicine reports, 20(4), 3027-3034 (2019-08-23)
Curcumin has been demonstrated to reduce markers of inflammation during acute pancreatitis (AP). However, the underlying mechanisms of the protective effects of curcumin are unknown. In the present study the effects of curcumin in an AP animal model and cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service