Skip to Content
Merck
All Photos(1)

Key Documents

SML3152

Sigma-Aldrich

MitoPQ

≥90% (HPLC)

Synonym(s):

1-Methyl-1′-[10′′-(triphenylphosphonio)decyl)dec-1"-yl]-4,4′-bipyridine-1,1′-diium triiodide, MPQ, Mito-PQ, MitoParaquat, Mitochondria-targeted paraquat, mtPQ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H46N2P·3I
CAS Number:
Molecular Weight:
954.48
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

powder

storage condition

desiccated

color

brown to brown-red

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

C[N+]1=CC=C(C=C1)C2=CC=[N+](C=C2)CCCCCCCCCC[P+](C3=CC=CC=C3)(C4=CC=CC=C4)C5=CC=CC=C5.[I-].[I-].[I-]

InChI

1S/C39H46N2P.3HI/c1-40-30-25-35(26-31-40)36-27-32-41(33-28-36)29-17-6-4-2-3-5-7-18-34-42(37-19-11-8-12-20-37,38-21-13-9-14-22-38)39-23-15-10-16-24-39;;;/h8-16,19-28,30-33H,2-7,17-18,29,34H2,1H3;3*1H/q+3;;;/p-3

InChI key

AOZZGHKENAZYTD-UHFFFAOYSA-K

General description

MitoPQ (MitoParaquat) is a mitochondria-targeted redox cycler composed of the lipophilic triphenylphosphonium (TP) cation-conjugated paraquat that induces mitochondria superoxide production via redox cycling at the complex I flavin site. MitoPQ increases mitochondria superoxide production at a ~1000-fold higher efficacy than untargeted paraquat using either live cells (fold of change of MitoSox fluorescence = 1.4/1 μM & 3.2/5 μM MitoPQ vs 1.2/5 mM PQ; C2C12 myoblasts) or isolated mitochondria (fold of increase of H2O2 efflux = 4.6/1 μM MitoPQ vs 2.6/1 mM PQ; rat heart mitochondria). MitoPQ is a valuable tool for conducting cellular and in vivo investigations into the role of mitochondrial superoxide generation in redox biology. Additionally, it can be utilized as a catalyst or co-stressor to replicate metabolic and neurodegenerative disease traits in experimental models.

Application

MitoPQ has been used to enhance mitochondrial reactive oxygen species (mitoROS) production in T-cell culture and investigate its influence over T-cell differentiation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bile acid metabolites control TH17 and Treg cell differentiation
Hang S, et al.
Nature, 143?148-143?148 (2019)
Ellen L Robb et al.
Free radical biology & medicine, 89, 883-894 (2015-10-11)
Superoxide is the proximal reactive oxygen species (ROS) produced by the mitochondrial respiratory chain and plays a major role in pathological oxidative stress and redox signaling. While there are tools to detect or decrease mitochondrial superoxide, none can rapidly and
Daniel J Fazakerley et al.
The Journal of biological chemistry, 293(19), 7315-7328 (2018-03-31)
Mitochondrial oxidative stress, mitochondrial dysfunction, or both have been implicated in insulin resistance. However, disentangling the individual roles of these processes in insulin resistance has been difficult because they often occur in tandem, and tools that selectively increase oxidant production
Eva Sidlauskaite et al.
Redox biology, 16, 344-351 (2018-03-28)
Developmental synapse pruning refines burgeoning connectomes. The basic mechanisms of mitochondrial reactive oxygen species (ROS) production suggest they select inactive synapses for pruning: whether they do so is unknown. To begin to unravel whether mitochondrial ROS regulate pruning, we made
Elizabeth C Hinchy et al.
The Journal of biological chemistry, 293(44), 17208-17217 (2018-09-21)
Mitochondrial reactive oxygen species (ROS) production is a tightly regulated redox signal that transmits information from the organelle to the cell. Other mitochondrial signals, such as ATP, are sensed by enzymes, including the key metabolic sensor and regulator, AMP-activated protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service