Skip to Content
Merck
All Photos(1)

Key Documents

SML2561

Sigma-Aldrich

ML162

≥98% (HPLC)

Synonym(s):

α-[(2-Chloroacetyl)(3-chloro-4-methoxyphenyl)amino]-N-(2-phenylethyl)-2-thiopheneacetamide, 2-Chloro-N-(3-chloro-4-methoxyphenyl)-N-(2-oxo-2-(phenethylamino)-1-(thiophen-2-yl)ethyl)acetamide, BRD-5421, BRD5421, CID 3689413, ML 162, ML-162, Molecular Libraries 162

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22Cl2N2O3S
CAS Number:
Molecular Weight:
477.40
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to very dark brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[s]1c(ccc1)C(N(c3cc(c(cc3)OC)Cl)C(=O)CCl)C(=O)NCCc2ccccc2

InChI

1S/C23H22Cl2N2O3S/c1-30-19-10-9-17(14-18(19)25)27(21(28)15-24)22(20-8-5-13-31-20)23(29)26-12-11-16-6-3-2-4-7-16/h2-10,13-14,22H,11-12,15H2,1H3,(H,26,29)

InChI key

UNVKYJSNMVDZJE-UHFFFAOYSA-N

Biochem/physiol Actions

Covalent phospholipid glutathione peroxidase (GPX4; GSHPx-4; PHGPx; snGPx; snPHGPx) inhibitor that induces ferroptosis.
ML162 is a small molecule that induces ferroptosis via covalent inhibition of cellular phospholipid glutathione peroxidase (GPX-4; GPX4; GSHPx-4; PHGPx; snGPx; snPHGPx). Synthetic lethality studies in cancer cultures identify contributing factors such as HRas(G12V) expression and fumarate hydratase (FH) deletion to ML162 sensitivity (IC50 = 25 nM & 35 nM, respectively, in BJeLR-HRas(G12V) & UOK262-FH-/- cultures).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michel Weïwer et al.
Bioorganic & medicinal chemistry letters, 22(4), 1822-1826 (2012-02-03)
Synthetic lethal screening is a chemical biology approach to identify small molecules that selectively kill oncogene-expressing cell lines with the goal of identifying pathways that provide specific targets against cancer cells. We performed a high-throughput screen of 303,282 compounds from
Emilie Logie et al.
International journal of molecular sciences, 22(22) (2021-11-28)
Disease relapse and therapy resistance remain key challenges in treating multiple myeloma. Underlying (epi-)mutational events can promote myelomagenesis and contribute to multi-drug and apoptosis resistance. Therefore, compounds inducing ferroptosis, a form of iron and lipid peroxidation-regulated cell death, are appealing
Vasanthi S Viswanathan et al.
Nature, 547(7664), 453-457 (2017-07-06)
Plasticity of the cell state has been proposed to drive resistance to multiple classes of cancer therapies, thereby limiting their effectiveness. A high-mesenchymal cell state observed in human tumours and cancer cell lines has been associated with resistance to multiple
Kenichi Shimada et al.
Cell chemical biology, 23(2), 225-235 (2016-02-09)
Precision medicine in oncology requires not only identification of cancer-associated mutations but also effective drugs for each cancer genotype, which is still a largely unsolved problem. One approach for the latter challenge has been large-scale testing of small molecules in
Qian Xue et al.
Autophagy, 19(7), 1982-1996 (2023-01-10)
Ferroptosis is a type of iron-dependent regulated cell death characterized by unrestricted lipid peroxidation and membrane damage. Although GPX4 (glutathione peroxidase 4) plays a master role in blocking ferroptosis by eliminating phospholipid hydroperoxides, the regulation of GPX4 remains poorly understood.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service