Skip to Content
Merck

SML3039

YKL-5-124

≥98% (HPLC)

Synonym(s):

(S)-3-(4-acrylamidobenzamido)-N-(2-(dimethylamino)-1-phenylethyl)-6,6-dimethyl-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxamide, N-[(1S)-2-(Dimethylamino)-1-phenylethyl]-4,6-dihydro-6,6-dimethyl-3-[[4-[(1-oxo-2-propen-1-yl)amino]benzoyl]amino]-pyrrolo[3,4-c]pyrazole-5(1H)-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H33N7O3
CAS Number:
Molecular Weight:
515.61
UNSPSC Code:
12352107
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN(C)C[C@@H](NC(N1CC(C(NC(C2=CC=C(NC(C=C)=O)C=C2)=O)=NN3)=C3C1(C)C)=O)C4=CC=CC=C4

InChI

1S/C28H33N7O3/c1-6-23(36)29-20-14-12-19(13-15-20)26(37)31-25-21-16-35(28(2,3)24(21)32-33-25)27(38)30-22(17-34(4)5)18-10-8-7-9-11-18/h6-15,22H,1,16-17H2,2-5H3,(H,29,36)(H,30,38)(H2,31,32,33,37)/t22-/m1/s1

InChI key

KPABJHHKKJIDGX-JOCHJYFZSA-N

Biochem/physiol Actions

YKL-5-124 is a potent and highly selective covalent inhibitor of cyclin-dependent kinase 7 (CDK7) that induces cell cycle arrest at the G1/S transition.
potent and highly selective covalent inhibitor of cyclin-dependent kinase 7 (CDK7)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library