Skip to Content
Merck
All Photos(1)

Key Documents

SML1392

Sigma-Aldrich

FDI-6

≥98% (HPLC)

Synonym(s):

3-Amino-N-(4-fluorophenyl)-6-(2-thienyl)-4-(trifluoromethyl)-thieno[2,3-b]pyridine-2-carboxamide, 3-Amino-N-(4-fluorophenyl)-6-(thiophen-2-yl)-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxamide, NCGC00099374

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H11F4N3OS2
CAS Number:
Molecular Weight:
437.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder (flocculent)

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=C(C(NC2=CC=C(F)C=C2)=O)SC3=C1C(C(F)(F)F)=CC(C4=CC=CS4)=N3

InChI

1S/C19H11F4N3OS2/c20-9-3-5-10(6-4-9)25-17(27)16-15(24)14-11(19(21,22)23)8-12(26-18(14)29-16)13-2-1-7-28-13/h1-8H,24H2,(H,25,27)

InChI key

ZATJMMZPGVDUOM-UHFFFAOYSA-N

Biochem/physiol Actions

FDI-6 is a potent and specific inhibitor of FOXM1 that blocks DNA binding. FDI-6 binds to FOXM1 protein and specifically downregulates FOXM1-activated genes.
Forkhead domain inhibitor-6 (FDI-6) is used to treat anaemia patients because of diminished red blood production. In Hep-2 cells, FDI-6 can stimulate cell death and also helps to prevent cell proliferation, invasion and migration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew P Sawaya et al.
Nature communications, 11(1), 4678-4678 (2020-09-18)
Diabetic foot ulcers (DFUs) are a life-threatening disease that often result in lower limb amputations and a shortened lifespan. However, molecular mechanisms contributing to the pathogenesis of DFUs remain poorly understood. We use next-generation sequencing to generate a human dataset
Susana Ros et al.
Cancer cell, 38(4), 516-533 (2020-09-26)
PIK3CA, encoding the PI3Kα isoform, is the most frequently mutated oncogene in estrogen receptor (ER)-positive breast cancer. Isoform-selective PI3K inhibitors are used clinically but intrinsic and acquired resistance limits their utility. Improved selection of patients that will benefit from these
Forkhead domain inhibitor-6 (FDI-6) increases apoptosis and inhibits invasion and migration of laryngeal carcinoma cells by down-regulating nuclear FoxM1
Liu Y, et al.
Chinese Journal of Catalysis, 33(5), 611-616 (2017)
Loss of Forkhead box M1 promotes erythropoiesis through increased proliferation of erythroid progenitors
Youn M, et al.
Haematologica, 102(5), 826-834 (2017)
Karan Ulhaka et al.
International journal of molecular sciences, 22(13) (2021-07-03)
Triple-negative breast cancer (TNBC) presents an important clinical challenge, as it does not respond to endocrine therapies or other available targeting agents. FOXM1, an oncogenic transcriptional factor, has reported to be upregulated and associated with poor clinical outcomes in TNBC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service