Skip to Content
Merck
All Photos(1)

Key Documents

SML1032

Sigma-Aldrich

KL001

≥98% (HPLC)

Synonym(s):

N-[3-(9H-Carbazol-9-yl)-2-hydroxypropyl]-N-(2-furanylmethyl)-methanesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22N2O4S
CAS Number:
Molecular Weight:
398.48
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N(CC(O)C[n]2c3c(c4c2cccc4)cccc3)Cc1[o]ccc1)C

InChI

1S/C21H22N2O4S/c1-28(25,26)22(15-17-7-6-12-27-17)13-16(24)14-23-20-10-4-2-8-18(20)19-9-3-5-11-21(19)23/h2-12,16,24H,13-15H2,1H3

InChI key

OQAFDLPAPSSOHY-UHFFFAOYSA-N

Biochem/physiol Actions

KL001 stabilizes cryptochrome (CRY), preventing ubiquitin-dependent degradation.
KL001 stabilizes cryptochrome (CRY), preventing ubiquitin-dependent degradation. CRY proteins are part of a feedback loop, acting as transcription repressors to inhibit CLOCK-BMAL1 components of the circadian clock. KL001 stabilization of CRY proteins results in lengthening of the circadian period, and also inhibited glucagon-induced gluconeogenesis in primary hepatocytes. Other compounds affecting circadian rhythms include Casein kinase I (CKI) inhibitors such as longdaysin (Sigma Prod. No. SML0127) and synthetic ligands for the nuclear receptors REV-ERB such as SR8278 (Sigma Prod. No. S9576), but KL001 appears to be the first small molecule that specifically acts on CRY proteins.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Alfred G Tamayo et al.
FEBS open bio, 10(9), 1868-1879 (2020-07-28)
Transcriptional dynamics of cancer cells govern cell fate decisions and are therapeutically actionable drug targets. In this study, we engineered a circulating cancer cell line that secretes a luciferase reporter to capture constitutive and circadian clock-driven transcription dynamics over the
Abdelhalim Azzi et al.
iScience, 26(8), 107449-107449 (2023-08-18)
Circadian clock controls daily behavior and physiology. The activity of various signaling pathways affects clock gene expression. Here, we show that the core circadian clock gene CRY1 is a direct target of the Hippo pathway effector YAP. YAP binds to
M Abdulhadi Alagha et al.
Cartilage, 13(2_suppl), 53S-67S (2020-02-16)
Circadian rhythms in cartilage homeostasis are hypothesized to temporally segregate and synchronize the activities of chondrocytes to different times of the day, and thus may provide an efficient mechanism by which articular cartilage can recover following physical activity. While the
Xiaodong Zhuang et al.
iScience, 24(10), 103144-103144 (2021-09-22)
The coronavirus disease 2019 pandemic, caused by severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) coronavirus, is a global health issue with unprecedented challenges for public health. SARS-CoV-2 primarily infects cells of the respiratory tract via spike glycoprotein binding to angiotensin-converting
Ayesha A Shafi et al.
Nature communications, 12(1), 401-401 (2021-01-17)
Mechanisms regulating DNA repair processes remain incompletely defined. Here, the circadian factor CRY1, an evolutionally conserved transcriptional coregulator, is identified as a tumor specific regulator of DNA repair. Key findings demonstrate that CRY1 expression is androgen-responsive and associates with poor

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service