Skip to Content
Merck
All Photos(1)

Key Documents

SML0679

Sigma-Aldrich

Danshensu sodium salt

≥98% (HPLC)

Synonym(s):

(R)-α,3,4-Trihydroxy-benzenepropanoic acid sodium salt, (R)-(+)-3-(3,4-Dihydroxyphenyl)lactic acid, 3-(3′,4′-Dihydroxyphenyl)-(2R)-lactic acid sodium salt, DS 182, Dan Shen Su, Dan shen suan A, Salianic acid A, Salvianic acid A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9O5 · Na
CAS Number:
Molecular Weight:
220.15
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +30 to +40°, c = 1 in H2O

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=C(O)C=CC(C[C@@H](O)C([O-])=O)=C1.[Na+]

InChI

1S/C9H10O5.Na/c10-6-2-1-5(3-7(6)11)4-8(12)9(13)14;/h1-3,8,10-12H,4H2,(H,13,14);/q;+1/p-1/t8-;/m1./s1

InChI key

ZMMKVDBZTXUHFO-DDWIOCJRSA-M

Biochem/physiol Actions

Danshensu is an active component of Salvia miltiorrhiza (Danshen) that suppresses the formation of reactive oxygen species, inhibits platelet adhesion and aggregation, and protects myocardium against ischemia. Danshensu is cardioprotective, which protects myocardium from the reperfusion injury and inhibits apoptosis of H9c2 cardiomyocytes via Akt and ERK1/2 phosphorylation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ye Jin et al.
Journal of pharmaceutical and biomedical analysis, 77, 32-39 (2013-01-30)
The application of near infrared (NIR) spectroscopy for on-line quantitative monitoring of alcohol precipitation of the Danhong injection was investigated. For the NIR measurements, two fiber optic probes designed to transmit NIR radiation through a 2mm path length flow cell
Jian-ping Zhang et al.
Planta medica, 79(9), 744-752 (2013-05-24)
The traditional Chinese medicinal herb Danshen (Salvia miltiorrhiza), first recorded in the "Shen Nong's Herbal Classic", has long been used to treat cardiovascular conditions, although the mechanism(s) underlying its effects remain unclear. Here, an iron dextran injection (50 mg ·
Libo Du et al.
Nanotoxicology, 7(3), 294-300 (2012-02-09)
To suppress the cytotoxicity of gold nanoparticles (AuNPs), danshensu, a naturally occurring polyphenol antioxidant isolated from Chinese herb, was used to provide a fundamental protection layer for AuNPs, to alleviate oxidative stress and as a reducing agent to react with
Qi-Tao Zhao et al.
Chinese journal of natural medicines, 10(1), 53-57 (2013-01-11)
To find out the role of salvianic acid A (SAA) in the protection of vascular endothelial cells (VEC) and its possible mechanism in vitro. The ingredient at various concentrations was added to human umbilical vein endothelial cells (HUVEC) treated with
Limin Zhou et al.
Journal of ethnopharmacology, 143(2), 648-655 (2012-08-08)
Danshen-Gegen (DG) product has clinically been proven to be an effective agent for heart-tonic efficacy by our previous research. In the mean time, herb-drug interactions between DG product and its commonly co-administered drugs, such as aspirin or warfarin need to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service