Skip to Content
Merck
All Photos(4)

Key Documents

C3878

Sigma-Aldrich

Chlorogenic acid

≥95% (titration)

Synonym(s):

CGA, 1,4,5-Trihydroxycyclohexanecarboxylic acid 3-(3,4-dihydroxycinnamate), 3-(3,4-Dihydroxycinnamoyl)quinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18O9
CAS Number:
Molecular Weight:
354.31
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥95% (titration)

form

powder

mp

210 °C (dec.) (lit.)

SMILES string

O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\c2ccc(O)c(O)c2)[C@@H]1O)C(O)=O

InChI

1S/C16H18O9/c17-9-3-1-8(5-10(9)18)2-4-13(20)25-12-7-16(24,15(22)23)6-11(19)14(12)21/h1-5,11-12,14,17-19,21,24H,6-7H2,(H,22,23)/b4-2+/t11-,12-,14-,16+/m1/s1

InChI key

CWVRJTMFETXNAD-JUHZACGLSA-N

Gene Information

rat ... G6pc(25634)

Looking for similar products? Visit Product Comparison Guide

General description

Chlorogenic acid is a bioactive polyphenolic compound mainly found in many plants[1], such as coffee.[2] It is a potent neuroprotectant[3] and also possesses antiviral[1], antifungal[3], antioxidant[2] and antitumor properties.[4] CGA may also be involved in regulating glucose and lipid metabolism and improving insulin sensitivity.[5]
Chlorogenic acid is a cinnamate ester used as a reducing agent and stabilizer in bimetallic nanoparticle synthesis.[6]

Application

  • Antioxidant and Phenolic Profiles: A study detailed the antioxidant and phenolic profiles of Thai medicinal plants, including chlorogenic acid, to support functional food and pharmaceutical development, highlighting its potential applications in enhancing food functionality and health benefits (Wanyo et al., 2024).
  • Intravesical Therapy for Cystitis: Research demonstrated the use of chlorogenic acid in intravesical therapy to modify acute voiding behavior in mice with systemic lipopolysaccharide inflammation-induced cystitis, suggesting therapeutic potential in bladder disorders (Yeh et al., 2024).
  • Cardioprotection in Chemotherapy: A hydroethanolic extract containing chlorogenic acid was investigated for its role in mitigating doxorubicin-induced cardiotoxicity through mitochondrial protection mediated by AMPK-PGC-1α-SOD, underscoring its potential in protective strategies against chemotherapy-induced cardiac damage (Song et al., 2024).
  • Nutritional Delivery and Food Industry: A review discussed chlorogenic acid′s role in nutrition delivery and its implications in the food industry, proposing its use as nanoparticles for active food packaging application. (Hu et al., 2024).

Caution

May yield a hazy solution in ethanol

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chlorogenic acid and caffeic acid are absorbed in humans
Olthof MR
The Journal of Nutrition, 131(1), 66-71 (2001)
Controlled green synthesis of Au-Pt bimetallic nanoparticles using chlorogenic acid
Chen R, et al.
Research on Chemical Intermediates, 47, 4051-4066 (2021)
Shp2 regulates chlorogenic acid-induced proliferation and adipogenic differentiation of bone marrow-derived mesenchymal stem cells in adipogenesis
Zhou RP
Molecular Medicine Reports, 11(6), 4489-4495 (2015)
Computational screen and experimental validation of anti-influenza effects of quercetin and chlorogenic acid from traditional Chinese medicine
Liu Z
Scientific Reports, 6, 19095-19095 (2016)
Chlorogenic acid and coffee prevent hypoxia-induced retinal degeneration
Jang H
Journal of Agricultural and Food Chemistry, 62(1), 182-191 (2013)

Questions

  1. Is this a synthetic product or is it isolated from natural sources?

    1 answer
    1. This product is isolated from natural sources. Please see the link below to review a sample Certificate of Origin:
      https://www.sigmaaldrich.com/coo/undefined/C3878/*

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service