Skip to Content
Merck
All Photos(1)

Key Documents

SML0366

Sigma-Aldrich

Cipamfylline

≥98% (HPLC)

Synonym(s):

8-Amino-1,3-bis(cyclopropylmethyl)-3,9-dihydro-1H-Purine-2,6-dione, BRL 61063

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17N5O2
CAS Number:
Molecular Weight:
275.31
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

Nc1nc2N(CC3CC3)C(=O)N(CC4CC4)C(=O)c2[nH]1

InChI

1S/C13H17N5O2/c14-12-15-9-10(16-12)17(5-7-1-2-7)13(20)18(11(9)19)6-8-3-4-8/h7-8H,1-6H2,(H3,14,15,16)

InChI key

KSPYMJJKQMWWNB-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

Cipamfylline is a PDE4 inhibitor that has been shown to cause a cellular redistribution of PDE4A4 into accretion foci, through an association with the ubiquitin scaffolding protein p62.
Cipamfylline is a PDE4 inhibitor.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Phosphodiesterases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R N Willette et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 17(2), 210-219 (1997-02-01)
The role of the phosphodiesterase type IV isozyme (PDE IV) in the regulation of cerebrovascular tone was investigated in the canine basilar artery in vitro and in vivo. The PDE isozymes extracted from the canine basilar artery were isolated by
David S Coombes et al.
Journal of pharmaceutical sciences, 91(7), 1652-1658 (2002-07-13)
We investigated the morphologies of three polymorphs of 1,3-di(cyclopropylmethyl)-8-aminoxanthine, a compound of pharmaceutical importance. We compared the experimental morphologies with those predicted by theoretical methods. We also predicted the elastic constants of the three polymorphs. These results are used to
D R Buckle et al.
Journal of medicinal chemistry, 37(4), 476-485 (1994-02-18)
Alkylation of the selective type IV phosphodiesterase inhibitor, 8-amino-1,3-bis(cyclopropylmethyl)-xanthine (1, BRL 61063), led exclusively to the N-7 substituted derivatives 2-9, which showed varying selectivities for the PDE type IV isoenzyme relative to PDE Va. The 4-methoxybenzyl derivative 6 in particular
A M Badger et al.
Circulatory shock, 44(4), 188-195 (1994-12-01)
Three inhibitors of calcium-dependent cyclic adenosine 3'5'-monophosphate (cAMP) dependent phosphodiesterase IV (PDE IV) were evaluated for their effects on lipopolysaccharide (LPS)-induced tumor necrosis factor (TNF) production in vitro and in vivo and for their ability to protect mice from LPS-induced
J M Kaplan et al.
Toxicology, 95(1-3), 187-196 (1995-01-06)
BRL 61063 is a novel xanthine phosphodiesterase (PDE) type IV inhibitor with selective inhibitory activity for tumor necrosis factor (TNF) alpha production. This compound inhibits TNF alpha production by activated human blood monocytes in vitro and in animal models of

Articles

Cyclic nucleotide phosphodiesterases (PDEs) catalyze the hydrolysis of cAMP and/or cGMP. There are 11 different mammalian PDE families.

Cyclic nucleotide phosphodiesterases (PDEs) catalyze the hydrolysis of cAMP and/or cGMP. There are 11 different mammalian PDE families.

Cyclic nucleotide phosphodiesterases (PDEs) catalyze the hydrolysis of cAMP and/or cGMP. There are 11 different mammalian PDE families.

Cyclic nucleotide phosphodiesterases (PDEs) catalyze the hydrolysis of cAMP and/or cGMP. There are 11 different mammalian PDE families.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service