Skip to Content
Merck

SML3673

Lin28 inhibitor C1632

≥98% (HPLC)

Synonym(s):

Lin28 1632, Lin28 inhibitor I, Lin28-C1632, Lin28-let-7 antagonist 1, N-Methyl-N-[3-(3-methyl[1,2,4]triazolo[4,3-b]pyridazin-6-yl)phenyl]acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H15N5O
CAS Number:
Molecular Weight:
281.31
UNSPSC Code:
12352200
NACRES:
NA.28
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

, White to light Brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[n]21nc(ccc2nnc1C)c3cc(ccc3)N(C)C(=O)C

InChI

1S/C15H15N5O/c1-10-16-17-15-8-7-14(18-20(10)15)12-5-4-6-13(9-12)19(3)11(2)21/h4-9H,1-3H3

InChI key

WPAQLESUVYGUJZ-UHFFFAOYSA-N

Biochem/physiol Actions

Compound 1632 (C1632) is a RNA-binding protein Lin28 inhibitor that prevents Lin28-mediated inhibition of let-7 microRNA (miRNA) maturation by blocking Lin28 interaction with let-7 miRNA precursor (IC50 = 8 μM by ELISA using truncated pre-let-7a-2). C1632 specifically upregulates Huh7 cellular let-7a/g/f miRNA, but not RNU44 snoRNA (60 μM for 48h), inhibits the stemness and induces differentiation of mouse embryonic stem cells (mESCs, 20 μM for 48h), as well as inhibits the proliferation and stem-like properties of human cancer stem cells (CSCs) in 22Rv1 and Huh7 cultures (IC50 20-42 μM).
RNA-binding protein Lin28 inhibitor that prevents Lin28-mediated inhibition of let-7 microRNA (miRNA) maturation.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library