Skip to Content
Merck
All Photos(2)

Key Documents

SAB5200110

Sigma-Aldrich

Anti-AQP2 antibody produced in rabbit

1 mg/mL, affinity isolated antibody

Synonym(s):

Anti-AQP-CD, Anti-AQP2, Anti-AQPCD, Anti-Aquaporin 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 28.8 kDa

species reactivity

human, mouse, rat

concentration

1 mg/mL

technique(s)

indirect immunofluorescence: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

rat ... Aqp2(25386)

General description

The gene ID of the protein aquaporin-2 (AQP2) is 25386.

Specificity

Detects ~28.8 kDa. May detect larger glycosylated bands ~35-50 kDa.

Immunogen

C-terminal peptide

Biochem/physiol Actions

Aquaporin 2 (AQP2) takes part in maintaining the water balance in cells. A high concentration of the protein has been observed in rats during hypothyroidism.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

PBS, pH 7..4, 50% glycerol, and 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorena I Sarati et al.
Metabolism: clinical and experimental, 62(9), 1287-1295 (2013-05-28)
Hypothyroid state and aging are associated with impairment in water reabsorption and changes in aquaporin water channel type 2 (AQP2). Nitric oxide (NO) is involved in AQP2 trafficking to the apical plasma membrane in medullary collecting duct cells. The purpose
Salvador Cazorla-Vázquez et al.
Cells, 12(15) (2023-08-11)
Adhesion G protein-coupled receptors (aGPCRs) comprise the second-largest class of GPCRs, the most common target for approved pharmacological therapies. aGPCRs play an important role in development and disease and have recently been associated with the kidney. Several aGPCRs are expressed
Peter Kösters et al.
Cells, 13(10) (2024-05-24)
Uncovering the function of understudied G protein-coupled receptors (GPCRs) provides a wealth of untapped therapeutic potential. The poorly understood adhesion GPCR Gpr126 (Adgrg6) is widely expressed in developing kidneys. In adulthood, Gpr126 expression is enriched in parietal epithelial cells (PECs)
William J Monis et al.
The Journal of cell biology, 216(7), 2131-2150 (2017-06-04)
Primary cilia perceive the extracellular environment through receptors localized in the ciliary membrane, but mechanisms directing specific proteins to this domain are poorly understood. To address this question, we knocked down proteins potentially important for ciliary membrane targeting and determined
Naohiro Nomura et al.
American journal of physiology. Cell physiology, 307(7), C597-C605 (2014-06-20)
A reduction or loss of plasma membrane aquaporin 2 (AQP2) in kidney principal cells due to defective vasopressin (VP) signaling through the VP receptor causes excessive urine production, i.e., diabetes insipidus. The amount of AQP2 on the plasma membrane is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service