Skip to Content
Merck
All Photos(1)

Key Documents

L0663

Sigma-Aldrich

Anti-Laminin-2 (α-2 Chain) antibody, Rat monoclonal

clone 4H8-2, purified from hybridoma cell culture

Synonym(s):

Anti-Merosin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4H8-2, monoclonal

form

buffered aqueous solution

mol wt

antigen ~400 kDa (denatured)

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry (frozen sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
indirect immunofluorescence: 4-8 μg/mL using acetone-fixed frozen sections of human tongue
microarray: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LAMA2(3908)
mouse ... Lama2(16773)

General description

The gene LAMA2 (laminin-2) encodes a protein belonging to a large family of heterotrimeric glycoproteins called laminins. It contains a large globular C-terminal domain that binds to dystroglycan and integrin α7β1 cell receptor specific for myofibers. The amino terminal domain associates with other laminins during the formation of protein meshwork that is involved in the basal membrane supramolecular assembly. The gene is mapped to human chromosome 6q22.33.
Laminin, the most abundant structural and biologically active component in basement membrane, is a complex extracellular glycoprotein of 700-900 kDa that plays an important role in many aspects of the cell biology. It is composed of one α chain (approx. 400 kDa, previously called A chain) one β chain (215 kDa, B1) and one γ chain (205 kDa, B2), held together by disulfide bonds.
Anti-Laminin-2 (α-2 Chain) antibody, Rat monoclonal (rat IgG1 isotype) is derived from the 4H8-2 hybridoma produced by the fusion of rat myeloma cells and splenocytes from a Lewis rat immunized with mouse heart laminin-2.1 The antibody is purified from the culture supernatant of hybridoma cells grown in a bioreactor. Anti-Laminin-2 (α-2 Chain) antibody, Rat monoclonal reacts specifically with mouse1 and human2, laminin-2. The epitope recognized by the antibody resides in the N-terminal portion of the α2 chain of laminin.

Specificity

The epitope recognized by the antibody resides in the N-terminal portion of the α2 chain of laminin.

Immunogen

Mouse heart laminin-2

Application

Anti-Laminin-2 (α-2 Chain) antibody, Rat monoclonal has been used in immunofluorescence and immunohistochemistry.

Biochem/physiol Actions

Laminins are glycoproteins that function in extracellular matrix architecture, cell adhesion, differentiation, and neurite outgrowth. Mutations in the gene LAMA2 (laminin-2) leading to either partial deficiency or complete absence of laminin α2 have been associated with limb-girdle muscular dystrophy (LGMD), characterized by severe, early-onset congenital muscular dystrophy to mild, later childhood-onset limb-girdle type muscular dystrophy, hypotonia with muscle weakness at early infancy, poor spontaneous movements with contractures of the large joints, and weak cry along with respiratory dysfunction.

Physical form

Solution in phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Preparation Note

Purified from the culture supernatant of hybridoma cells grown in a bioreactor.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in "frostfree" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of pericytic laminin in blood brain barrier integrity maintenance.
Gautam J, et.al.
Scientific Reports, 6, 36450-36450 (2016)
Expression and distribution of laminin a1 and a2 chains in embryonic and adult mouse tissues: an immunochemical approach.
Sasaki T, et al.
Experimental Cell Research, 275(2), 185-199 (2002)
Nicole D Paris et al.
eLife, 5 (2016-11-20)
Skeletal muscle regenerative potential declines with age, in part due to deficiencies in resident stem cells (satellite cells, SCs) and derived myogenic progenitors (MPs); however, the factors responsible for this decline remain obscure. TGFβ superfamily signaling is an inhibitor of
Cathepsin S Contributes to the Pathogenesis of Muscular Dystrophy in Mice.
Tjondrokoesoemo A
The Journal of Biological Chemistry, 291, 9920-9928 (2016)
Thomas C Roberts et al.
Molecular therapy. Nucleic acids, 1, e39-e39 (2013-01-25)
MicroRNAs (miRNAs) are a class of small RNAs that regulate gene expression and are implicated in wide-ranging cellular processes and pathological conditions including Duchenne muscular dystrophy (DMD). We have compared differential miRNA expression in proximal and distal limb muscles, diaphragm

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service