Skip to Content
Merck
All Photos(2)

Key Documents

S8753

Sigma-Aldrich

Strychnine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22N2O2 · HCl
CAS Number:
Molecular Weight:
370.87
EC Number:
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.32

SMILES string

Cl.[H][C@@]12CC(=O)N3c4ccccc4[C@]56CCN7CC(=CCO1)[C@]([H])(C[C@@]57[H])[C@]2([H])[C@]36[H]

InChI

1S/C21H22N2O2.ClH/c24-18-10-16-19-13-9-17-21(6-7-22(17)11-12(13)5-8-25-16)14-3-1-2-4-15(14)23(18)20(19)21;/h1-5,13,16-17,19-20H,6-11H2;1H/t13-,16-,17-,19-,20-,21+;/m0./s1

InChI key

VLXYTKMPCOQKEM-ZEYGOCRCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Strychnine hydrochloride has been used as a glycine receptor antagonist in the retina for biophysiological functions. It has also been used to block glutamatergic, GABAergic and glycinergic receptors and gap junctions.

Biochem/physiol Actions

Convulsant; glycine receptor antagonist not associated with the NMDA receptor.
Strychnine is a potent convulsant, which is extracted from Strychnos nux vomica seeds. At high doses, the compound causes convulsions and ultimately death due to respiratory paralysis. Strychnine acts as a respiratory, circulatory and digestive stimulant.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biomarkers in Toxicology null
Horizontal cell feedback without cone type-selective inhibition mediates red?green color opponency in midget ganglion cells of the primate retina
Crook JD, et al.
The Journal of Neuroscience, 31(5), 1762-1772 (2011)
Direct activation of the Mauthner cell by electric field pulses drives ultrarapid escape responses
Tabor KM, et al.
Journal of Neurophysiology, 112(4), 834-844 (2014)
Y-cell receptive field and collicular projection of parasol ganglion cells in macaque monkey retina
Crook JD, et al.
The Journal of Neuroscience, 28(44), 11277-11291 (2008)
Strychnos nux-vomica seeds: Pharmacognostical standardization, extraction, and antidiabetic activity
Bhati R, et al.
Journal of Ayurveda and integrative medicine, 3(2), 80-80 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service