Skip to Content
Merck
All Photos(3)

Key Documents

P1675

Sigma-Aldrich

Picrotoxin

powder

Synonym(s):

Cocculin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H34O13
CAS Number:
Molecular Weight:
602.58
Beilstein:
3894406
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

color

white

mp

203 °C (lit.)

SMILES string

[H][C@@]12C[C@@]3(O)[C@@]4([H])[C@H](C(C)=C)[C@]([H])(OC4=O)[C@@]5([H])OC(=O)[C@]1(O2)[C@@]35C.[H][C@@]67C[C@@]8(O)[C@@]9([H])[C@@H]([C@]([H])(OC9=O)[C@@]%10([H])OC(=O)[C@]6(O7)[C@@]8%10C)C(C)(C)O

InChI

1S/C15H18O7.C15H16O6/c1-12(2,18)6-7-10(16)20-8(6)9-13(3)14(7,19)4-5-15(13,22-5)11(17)21-9;1-5(2)7-8-11(16)19-9(7)10-13(3)14(8,18)4-6-15(13,21-6)12(17)20-10/h5-9,18-19H,4H2,1-3H3;6-10,18H,1,4H2,2-3H3/t5-,6+,7+,8+,9-,13-,14-,15+;6-,7+,8+,9+,10-,13-,14-,15+/m11/s1

InChI key

VJKUPQSHOVKBCO-NGKRNLQBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Picrotoxin is isolated from moonseed family, Menispermaceae. It is a stimulant and may cause convulsions. It is an allosteric modulator of γ-aminobutyric acid (GABAA) receptor.

Application

Picrotoxin has been used:
  • as non-competitive antagonist for the γ-aminobutyric acid (GABAA) for studying synchronized burst (SB) dynamics
  • as a component of extracellular saline for blocking glutamate and acetylcholine receptors in neurons
  • as a component of artificial cerebrospinal fluid (ACSF) for voltage-clamp recordings studies in dissociated cortical neurons

Biochem/physiol Actions

GABAA receptor antagonist; binds to the GABA receptor-linked Cl channel.
Picrotoxin is a competitive inhibitor of glycine receptors (GlyRs). It is a non-competitive antagonist, which binds to ligand bounded γ-aminobutyric acid (GABAA) receptor.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the GABAA Receptors and GABAC Receptors pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Other Notes

Molecular complex (1:1) of picrotin and picrotoxinin.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dynamic transitions among multiple oscillators of synchronized bursts in cultured neural networks
Kim JH, et al.
Journal of Statistical Mechanics: Theory and Experiment, 2014(4), P04019-P04019 (2014)
Aric C Madayag et al.
Brain structure & function, 224(7), 2311-2324 (2019-06-16)
Despite evidence that morphine-related pathologies reflect adaptations in NAc glutamate signaling, substantial gaps in basic information remain. The current study examines the impact of non-contingent acute, repeated, and withdrawal-inducing morphine dosing regimens on glutamate transmission in D1- or D2-MSNs in
Picrotoxin-like channel blockers of GABAA receptors
Olsen RW
Proceedings of the National Academy of Sciences of the USA, 103(16), 6081-6082 (2006)
On the mechanism of action of picrotoxin on GABA receptor channels in dissociated sympathetic neurones of the rat
Newland CF and Cull-Candy SG
The Journal of Physiology, 447(1), 191-213 (1992)
Andrew R Rau et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 39(33), 6449-6459 (2019-06-27)
Neuronal circuits regulating hunger and satiety synthesize information encoding the energy state of the animal and translate those signals into motivated behaviors to meet homeostatic needs. Proopiomelanocortin (POMC) neurons in the arcuate nucleus of the hypothalamus are activated by energy

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service