Skip to Content
Merck
All Photos(1)

Documents

S1799

Sigma-Aldrich

SP Sepharose

Fast Flow, aqueous ethanol suspension, 45-165 μm (wet), exclusion limit ~4,000,000 Da

Synonym(s):

Sulfopropyl-Sepharose

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
47101511
NACRES:
NA.56

Quality Level

form

aqueous ethanol suspension

matrix active group

, —CH2-SO3-

particle size

45-165 μm (wet)

pore size

~4,000,000 Da exclusion limit

capacity

180-250 μeq/mL, gel

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

SP Sepharose is used in protein chromatography, ion exchange chromatography and cation exchange media. SP Sepharose has been used to study inhibitory proteins and peptides from the rhizomes of zingiberaceae plants as well as to study the subunit heterogeneity and molecular evolution of soybean basic 7S globulin. SP Sepharose has also assisted with advancing industrial applications of preparation of chitosan oligosaccharides.

Physical form

Suspension in 0.2 M sodium acetate and 20% ethanol

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 - 109.4 °F

Flash Point(C)

38 - 43 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Güntert et al.
Journal of Alzheimer's disease : JAD, 21(2), 585-596 (2010-06-24)
Alzheimer's disease (AD) is a progressive neurodegenerative disorder where definite diagnosis can only be made postmortem, and for which the most promising peripheral markers of disease state and severity have been found in the cerebrospinal fluid. However, recent results suggest
Maneerat Yodjun et al.
Applied biochemistry and biotechnology, 166(8), 2037-2050 (2012-03-07)
Ammonium sulphate cut protein extracts, and their pepsin hydrolysates, from the rhizomes of 15 plants in the Zingiberaceae family were screened for their in vitro angiotensin I-converting enzyme inhibitory (ACEI) activity. The protein extract from Zingiber ottensii had the highest
Andrei D Shutov et al.
Bioscience, biotechnology, and biochemistry, 74(8), 1631-1634 (2010-08-12)
Basic 7S globulin, a cysteine-rich protein from soybean seeds, consists of subunits containing 27 kD and 16 kD chains linked by disulfide bonding. Three differently sized subunits of the basic 7S globulin were detected and partially separated by SP Sepharose
C Y Cheng et al.
Biotechnology and applied biochemistry, 32 ( Pt 3), 197-203 (2000-12-15)
A chitosan-degrading fungus, designated Aspergillus sp. Y2K, was isolated from soil. The micro-organism was used for producing chitosanase (EC 3.2.1.132) in a minimal medium containing chitosan as the sole carbon source. The induced chitosanase was purified to homogeneity from the
James C Geoghegan et al.
The Journal of biological chemistry, 282(50), 36341-36353 (2007-10-18)
The central pathogenic event of prion disease is the conformational conversion of a host protein, PrPC, into a pathogenic isoform, PrPSc. We previously showed that the protein misfolding cyclic amplification (PMCA) technique can be used to form infectious prion molecules

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service