Skip to Content
Merck
All Photos(1)

Key Documents

S1629

Sigma-Aldrich

Sulfatase from Aerobacter aerogenes

Type VI, buffered aqueous glycerol solution, 2-5 units/mg protein (biuret), 10-20 units/mL

Synonym(s):

Aryl-sulfatase, Aryl-sulfate sulfohydrolase, Phenolsulfatase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

type

Type VI

Quality Level

form

buffered aqueous glycerol solution

specific activity

2-5 units/mg protein (biuret)

mol wt

~41 kDa

concentration

10-20 units/mL

foreign activity

β-Glucuronidase ≤10 U/mL

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Sulfatases comprise Cys/Ser-X-Pro-X-Arg motif that is conserved in their active sites.

Application

Sulfatase from Aerobacter aerogenes has been used:
  • as a deconjugation enzyme for treating plasma samples for quercetin quantification using liquid chromatography with tandem mass spectrometry (LC/MS/MS) analyses
  • in fluorescence intensity-based enzymatic assay with an activity-based probe probe 1
  • to treat sulfatide liposomes to remove the 3-O-sulfogalactosyde head from sulfatides

Biochem/physiol Actions

Sulfatases hydrolyze sulfate ester bonds to generate inorganic sulfates. Microbial sulfatases participate in sulfur scavenging and are essential enzymes for the utilization of sulfur. They may be associated with pathogenesis. Commercially available sulfatase from Aerobacter aerogenes is useful as a deconjugation enzyme for the removal of glucuronate and sulfate moieties from conjugates. Sulfatases find application in industry and agriculture.

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrophenyl sulfate per min at pH 7.1 at 37 °C.

Physical form

Solution in 50% glycerol containing 0.01 M Tris, pH 7.5.

substrate

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melanie Glauser et al.
Clinica chimica acta; international journal of clinical chemistry, 430, 125-128 (2014-01-15)
Total (i.e. free+sulfated) metanephrines in plasma is a biomarker for the diagnosis of pheochromocytoma/paraganglioma. Sulfated metanephrines must be completely deconjugated by perchloric acid hydrolysis or sulfatase treatment prior to analytical measurement to enable quantification by current techniques. In this report
T Saidha et al.
Archives of biochemistry and biophysics, 272(1), 237-244 (1989-07-01)
Mitochondria that have been purified from cells of light-grown wild-type Euglena gracilis Klebs var. bacillaris Cori or dark-grown mutant W10BSmL and incubated with 35SO4(2-) and ATP accumulate a labeled compound in the surrounding medium. This compound is also labeled when
Toshiyuki Nakamura et al.
Bioscience, biotechnology, and biochemistry, 75(8), 1506-1510 (2011-08-09)
β-Glucuronidase and sulfatase are the major deconjugating enzymes used in the cleavage of the glucuronate and sulfate moieties, respectively, from certain conjugated food factors including polyphenols. In the present study, we found that compounds having the same molecular weights as
C Gil et al.
Biochimica et biophysica acta. Biomembranes, 1861(1), 161-169 (2018-11-23)
Epsilon toxin (Etx) from Clostridium perfringens is synthesized as a very low-active prototoxin form (proEtx) that becomes active upon proteolytic activation and has the capacity to cross the blood-brain barrier (BBB), thereby producing severe neurological effects. The identity and requirements
Toyokazu Miura et al.
Bioscience, biotechnology, and biochemistry, 70(6), 1509-1512 (2006-06-24)
Arylsulfatase activity was detected in a bacterial strain, Citrobacter braakii 69-b, isolated from soil by enrichment cultivation using porcine gastric mucin. The production of arylsulfatase was derepressed markedly in a synthetic medium by the addition of tyramine. The purified enzyme

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service