Skip to Content
Merck

Skip To

G7396

β-Glucuronidase from Escherichia coli

Type IX-A, lyophilized powder, 1,000,000-5,000,000 units/g protein (30 min assay)

Synonym(s):

β-D-Glucuronide glucuronosohydrolase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-606-8
MDL number:
EC Number:
Specific activity:
1,000,000-5,000,000 units/g protein (30 min assay)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


type

Type IX-A

Quality Segment

form

lyophilized powder

specific activity

1,000,000-5,000,000 units/g protein (30 min assay)

mol wt

290 kDa

composition

Protein, 30-70% biuret

shipped in

wet ice

storage temp.

−20°C

Application

New Technical Article Comparing Performance of Different Enzymes
Learn more
about recent application data generated by Sigma R&D to optimize hydrolysis for different drug classes using enzymes from different sources and the use of a chromatographicaly purified enzyme to reduce the effect of esterase activity resulting in conversion of 6-MAM to Morphine
Effective in the hydrolysis of steroid glucuronides.
Used for the hydrolysis of glucuronide conjugates in urinary metabolite analysis

Analysis Note

Contains buffer salts and stabilizer.

Other Notes

One Sigma or modified Fishman unit will liberate 1.0 μg of phenolphthalein from phenolphthalein glucuronide per hr at 37 °C at the pH 6.8 (30 min assay).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G7646G8295G8162
specific activity

1,000,000-5,000,000 units/g protein (30 min assay)

specific activity

5,000,000-20,000,000 units/g protein, pH 6.8 (30 min assay)

specific activity

≥10,000,000 units/g protein (30 min assay)

specific activity

≥5,000,000 units/g protein, pH 6.8 (biuret)

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

aqueous glycerol solution

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

shipped in

wet ice

shipped in

-

shipped in

-

shipped in

wet ice

mol wt

290 kDa

mol wt

290 kDa

mol wt

~290 kDa

mol wt

69-71 kDa

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

200


Still not finding the right product?


pictograms

Health hazard

signalword

Danger

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)

hcodes

Hazard Classifications

Resp. Sens. 1

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters