Skip to Content
Merck
All Photos(1)

Key Documents

RAB1138

Sigma-Aldrich

Rat BDNF ELISA Kit

for cell culture supernatants, plasma, and serum samples

Synonym(s):

BDNF, Brain-derived neurotrophic factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human, rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum sample(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 12 pg/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... Bdnf(24225)

General description

The brain-derived neurotrophic factor (BDNF) gene is mapped to human chromosome 11p14.1. The gene encodes a precursor protein, proBDNF. Mature BDNF (mBDNF) is synthesized by post-translational cleavage of proBDNF.
This ELISA antibody pair detects rat and human BDNF. Other species are not determined.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Biochem/physiol Actions

Brain-derived neurotrophic factor (BDNF) is involved in hippocampal function and verbal episodic memory in humans. BDNF also plays a vital role in vascular function and participates in angiogenesis via the specific receptor tropomyosin-related kinase B (TrkB). It is involved in the pathogenesis of Alzheimer′s disease.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Kang et al.
Brain sciences, 11(1) (2020-12-31)
(1) Background: Depression is one of the overwhelming public health problems. Alleviating hippocampus injury may prevent depression development. Herein, we established the chronic unpredictable mild stress (CUMS) model and aimed to investigate whether aerobic exercise (AE) could alleviate CUMS induced
The BDNF val66met polymorphism affects activity-dependent secretion of BDNF and human memory and hippocampal function.
Egan MF, et al.
Cell, 112(2), 257-269 (2003)
Peripheral vascular reactivity and serum BDNF responses to aerobic training are impaired by the BDNF Val66Met polymorphism.
Lemos JR Jr, et al.
Physiological Genomics, 48(2), 116-123 (2015)
Irina Naletova et al.
International journal of molecular sciences, 22(24) (2021-12-25)
l-carnosine (β-alanyl-l-histidine) (Car hereafter) is a natural dipeptide widely distributed in mammalian tissues and reaching high concentrations (0.7-2.0 mM) in the brain. The molecular features of the dipeptide underlie the antioxidant, anti-aggregating and metal chelating ability showed in a large
11p14. 1 microdeletions associated with ADHD, autism, developmental delay, and obesity.
Shinawi M, et al.
American Journal of Medical Genetics. Part A, 155(6), 1272-1280 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service