Skip to Content
Merck
All Photos(1)

Documents

RAB0707

Sigma-Aldrich

Human LDLR / Low-density Lipoprotein Receptor ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 8 pg/mL
standard curve range: 8.19-2000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... LDLR(3949)

General description

The antibody pair provided in this kit recognizes human Low-density Lipoprotein Receptor.
LDLR (low density lipoprotein receptor) codes for a transmembrane receptor glycoprotein, containing 860 amino acids and is mapped to human chromosome 19p13.2.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

LDLR′s (low density lipoprotein receptor) major function is to regulate the level of lipoprotein particles by mediating the clearance of LDL-cholesterol and triglyceride through receptor mediated endocytosis. LDLR is involved in proliferation by providing cholesterol for membrane synthesis. The LDLR gene is a target for treating cardiovascular diseases. Mutation in the gene causes familial hypercholesterolemia.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel S Hsia et al.
The British journal of nutrition, 124(6), 577-585 (2020-04-18)
Cranberries are high in polyphenols, and epidemiological studies have shown that a high-polyphenol diet may reduce risk factors for diabetes and CVD. The present study aimed to determine if short-term cranberry beverage consumption would improve insulin sensitivity and other cardiovascular
5-Azacytidine engages an IRE1?-EGFR-ERK1/2 signaling pathway that stabilizes the LDL receptor mRNA.
Mnasri N, et al.
Biochimica et Biophysica Acta - Gene Regulatory Mechanisms, 1861(1), 29-40 (2018)
Mutational screening in the LDLR gene among patients presenting familial hypercholesterolemia in the Southeast of Brazil.
Molfetta G A, et al.
Genetics and molecular research : GMR, 16(3) (2017)
E Soemirien Kasanmoentalib et al.
Journal of neuroinflammation, 16(1), 279-279 (2019-12-31)
The complement system is a vital component of the inflammatory response occurring during bacterial meningitis. Blocking the complement system was shown to improve the outcome of experimental pneumococcal meningitis. Complement factor H (FH) is a complement regulatory protein inhibiting alternative
A novel indel variant in LDLR responsible for familial hypercholesterolemia in a Chinese family.
Shu H, et al.
PLoS ONE, 12(12), e0189316-e0189316 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service