Skip to Content
Merck
All Photos(1)

Key Documents

RAB0661

Sigma-Aldrich

Human LGALS3 / Galectin-3 ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.6 ng/mL
standard curve range: 0.819-200 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... LGALS3(3958)

General description

The LGALS3 enzyme-linked immunosorbent assay (ELISA) kit provides for the quantitative measurement of galectin-3 in cell culture supernatants, plasma, and serum. Galectin-3 belongs to the β-galactoside-binding protein family. It is an intracellular and extracellular lectin. Galectin-3 is expressed in immune cells and epithelial cells. It interacts with intracellular glycoproteins and plays a role in several biological processes such as angiogenesis, apoptosis, cell growth, adhesion, and differentiation.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oliver Dörr et al.
Clinical cardiology, 41(9), 1164-1169 (2018-06-14)
Percutaneous mitral valve repair (PMVR) is an interventional treatment option in patients with severe mitral regurgitation (MR) and at high risk for open-heart surgery. Currently, limited information exists about predictors of procedural success after PMVR. Galectin-3 (Gal-3) and suppression of
My-Nhan Nguyen et al.
Scientific reports, 8(1), 8213-8213 (2018-05-31)
Galectin-3 is a biomarker of heart disease. However, it remains unknown whether increase in galectin-3 levels is dependent on aetiology or disease-associated conditions and whether diseased heart releases galectin-3 into the circulation. We explored these questions in mouse models of
Haibing Sun et al.
Critical care (London, England), 25(1), 109-109 (2021-03-20)
Galectin-3 (Gal-3) is a pleiotropic glycan-binding protein shown to be involved in sepsis and acute kidney injury (AKI). However, its role has never been elucidated in sepsis-associated AKI (S-AKI). We aimed to explore Gal-3's role and its potential utility as
Prasoon Verma et al.
Pediatric research, 85(4), 527-532 (2019-01-27)
Invasive candidiasis is an important cause of fungal infections in immunocompromised patients, including premature infants. The S-type lectin, galectin-3 (gal3), is increasingly recognized for its role in antifungal host defense. This study tested the hypothesis that tissue gal3 expression is
Yukinori Takenaka et al.
Glycoconjugate journal, 19(7-9), 543-549 (2004-02-06)
Galectin-3, a 31 kDa member of the beta-galactoside-binding proteins, is an intracellular and extracellular lectin which interacts with intracellular glycoproteins, cell surface molecules and extracellular matrix proteins. Galectin-3 is expressed widely in epithelial and immune cells and its expression is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service