Skip to Content
Merck
All Photos(3)

Documents

R8759

Sigma-Aldrich

Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae)

Type X-SA, lyophilized powder

Synonym(s):

Transfer RNA, tRNA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.51

type

Type X-SA

Quality Level

form

lyophilized powder

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Ribonucleic acid, transfer from baker′s yeast S. cerevisiae may be used as a starting material for the purification of specific amino acyl-tRNA species by methods such as liquid column chromatography or high-performance liquid chromatography (HPLC).

Unit Definition

One unit will yield an A260 of 1.0 in 1.0 mL of water (1 cm light path).

Preparation Note

Prepared by a modification of the procedure of Hancher, C.W., et al., Biotechnol. Bioeng., 11, 1055 (1969).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Maureen A Sartor et al.
Nucleic acids research, 34(1), 185-200 (2006-01-07)
The recent sequencing of a large number of Xenopus tropicalis expressed sequences has allowed development of a high-throughput approach to study Xenopus global RNA gene expression. We examined the global gene expression similarities and differences between the historically significant Xenopus
Ofri Levi et al.
Nucleic acids research, 49(1), 432-443 (2020-12-12)
Modification of nucleotides within an mRNA emerges as a key path for gene expression regulation. Pseudouridine is one of the most common RNA modifications; however, only a few mRNA modifiers have been identified to date, and no one mRNA pseudouridine
Violaine I Mottier-Pavie et al.
Developmental biology, 417(1), 50-62 (2016-07-02)
The mechanisms that modulate and limit the signaling output of adult stem cell niches remain poorly understood. To gain further insights into how these microenvironments are regulated in vivo, we performed a candidate gene screen designed to identify factors that
Jaclyn M Fingerhut et al.
PLoS genetics, 15(5), e1008028-e1008028 (2019-05-10)
Intron gigantism, where genes contain megabase-sized introns, is observed across species, yet little is known about its purpose or regulation. Here we identify a unique gene expression program utilized for the proper expression of genes with intron gigantism. We find
Peter Kozulin et al.
Molecular vision, 15, 2649-2662 (2009-12-17)
Recently we identified high levels of expression of Eph-A6 in the macula of developing human retina and showed localization of Eph-A6 to ganglion cells (GC). In the present study we investigated the expression of some members of the ephrin family

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service