Skip to Content
Merck
All Photos(3)

Key Documents

R7876

Sigma-Aldrich

Ribonucleic acid, transfer from wheat germ

Type V, 15-19 units/mg solid

Synonym(s):

Transfer RNA, tRNA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.55
Pricing and availability is not currently available.

grade

for molecular biology

Quality Level

type

Type V

form

lyophilized powder

specific activity

15-19 units/mg solid

storage condition

desiccated

solubility

H2O: ~2 mg/mL, clear to hazy

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Wheat germ tRNA contains many different specific tRNA molecules.

Application

Crude transfer RNA (tRNA), purified from wheat germ, contains many specific tRNA molecules that can be separated by different processes. Experiments for detailed investigation of structure and function using in vitro translation systems often require highly purified tRNA, which can be achieved by combinations of chromatographic separations and reversible biochemical modification of the tRNA. Wheat germ tRNA has been used for the purification of RNAse Z (tRNA 3′- processing endonucleases) from wheat germ. The purification included six steps with the most efficient one using an affinity column made of wheat germ tRNA, which the RNAse Z bound tightly.
Suitable for use as a carrier in nucleic acid purification and precipitation.

Components

tRNA is provided as a lyophilized powder that is soluble in water at a concentration of 2 mg/ml.

Unit Definition

One unit will yield an absorbance of 1.0 at 260 nm in 1.0 mL of water (1 cm light path).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spatial and temporal expression of the zebrafish genome by large-scale in situ hybridization screening.
Bernard Thisse et al.
Methods in cell biology, 77, 505-519 (2004-12-18)
Christèle Maison et al.
Nature genetics, 30(3), 329-334 (2002-02-19)
Post-translational modification of histone tails is thought to modulate higher-order chromatin structure. Combinations of modifications including acetylation, phosphorylation and methylation have been proposed to provide marks recognized by specific proteins. This is exemplified, in both mammalian cells and fission yeast
Yana P Blokhina et al.
PLoS genetics, 15(1), e1007730-e1007730 (2019-01-18)
Meiosis is a cellular program that generates haploid gametes for sexual reproduction. While chromosome events that contribute to reducing ploidy (homologous chromosome pairing, synapsis, and recombination) are well conserved, their execution varies across species and even between sexes of the
Sichen Shao et al.
Molecular cell, 50(5), 637-648 (2013-05-21)
Quality control of defective mRNAs relies on their translation to detect the lesion. Aberrant proteins are therefore an obligate byproduct of mRNA surveillance and must be degraded to avoid disrupting protein homeostasis. These defective translation products are thought to be
Dawid Krokowski et al.
The Journal of biological chemistry, 288(24), 17202-17213 (2013-05-07)
Endoplasmic reticulum (ER) stress-induced responses are associated with the loss of insulin-producing β-cells in type 2 diabetes mellitus. β-Cell survival during ER stress is believed to depend on decreased protein synthesis rates that are mediated via phosphorylation of the translation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service