Skip to Content
Merck
All Photos(1)

Documents

R8279

Sigma-Aldrich

RG108

≥98% (HPLC), powder

Synonym(s):

N-Phthalyl-L-tryptophan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14N2O4
CAS Number:
Molecular Weight:
334.33
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -250 to -310°, c = 0.5 in methanol

color

yellow

solubility

DMSO: >10 mg/mL

storage temp.

−20°C

SMILES string

OC(=O)[C@H](Cc1c[nH]c2ccccc12)N3C(=O)C4=CCCC=C4C3=O

InChI

1S/C19H16N2O4/c22-17-13-6-1-2-7-14(13)18(23)21(17)16(19(24)25)9-11-10-20-15-8-4-3-5-12(11)15/h3-8,10,16,20H,1-2,9H2,(H,24,25)/t16-/m0/s1

InChI key

LMAZUAXDZRILNJ-INIZCTEOSA-N

General description

RG108, a non-nucleoside analog is also called [2-(1,3-dioxoisoindolin-2-yl)-3-(1H-indol-3-yl) propanoic acid.

Application

RG108 has been used:
  • in reprogramming and increasing cell plasticity of primary multipotent mesenchymal stromal cells (MMSC)
  • as an inhibitor of DNA methyltransferase (DNMTi) in human retinal pigment epithelial ARPE-19 cells
  • as a DNA methyltransferase inhibitor in C33A2 cells to test its effect on papillomavirus late gene expression (HPV16)

Biochem/physiol Actions

RG108 is a DNA methyltransferase (DMNT) inhibitor. It reactivates tumor suppressor gene expression (p16, SFRP1, secreted frizzled related protein-1, and TIMP-3) in tumor cells by DNA demethylation. RG108 also inhibits human tumor cell line (HCT116, NALM-6) proliferation and increased doubling time in culture.
RG108 is less cytotoxic compared to azacytidine (5-Aza-dCR). It mediates radiosensitivity in esophageal cancer cells (EC). RG108 is a potent epigenetic modulator and a demethylating agent. It elicits non-cytotoxic functionality and may be useful in stem cell therapies based on human bone marrow-derived mesenchymal stem cells (hBMSCs). It interacts with DNA methyltransferase 1 at the catalytic domain region.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qing Wang et al.
Journal of cellular biochemistry, 119(2), 1501-1510 (2017-08-05)
DNA methylation plays a crucial role in lots of biological processes and cancer. 5-azacytidine (5-AC), a DNA methylation inhibitor, has been used as a potential chemotherapeutic agent for cancer. In this study, we used 5-AC treatment to investigate whether DNA
Paulina Tokarz et al.
European journal of pharmacology, 776, 167-175 (2016-02-24)
Epigenetic modifications influence DNA damage response (DDR). In this study we explored the role of DNA methylation and histone acetylation in DDR in cells challenged with acute or chronic oxidative stress. We used retinal pigment epithelial cells (ARPE-19), which natively
Yao Ou et al.
Oncology reports, 39(3), 993-1002 (2018-01-13)
Esophageal cancer (EC) is the eighth most common highly aggressive cancer worldwide. The purpose of this study was to investigate the effect of the DNA methyltransferase inhibitor RG108 on the radiosensitivity of EC cells. MTT and clonogenic assays were performed
Cecilia Johansson et al.
Virology, 482, 244-259 (2015-04-23)
We report that many histone modifications are unevenly distributed over the HPV16 genome in cervical cancer cells as well as in HPV16-immortalized keratinocytes. For example, H3K36me3 and H3K9Ac that are common in highly expressed cellular genes and over exons, were
Lucia Bialesova et al.
Scientific reports, 7(1), 10152-10152 (2017-09-02)
Increased adipocyte lipolysis links obesity to insulin resistance. The lipid droplet coating-protein Perilipin participates in regulation of lipolysis and is implicated in obesity. In the present study we investigate epigenetic regulation of the PLIN1 gene by correlating PLIN1 CpG methylation

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service